MilliporeSigma
  • LncRNA-N1LR Enhances Neuroprotection Against Ischemic Stroke Probably by Inhibiting p53 Phosphorylation.
LncRNA-N1LR Enhances Neuroprotection Against Ischemic Stroke Probably by Inhibiting p53 Phosphorylation.
Molecular neurobiology (2016-11-16)
Zhuomin Wu, Ping Wu, Xialin Zuo, Na Yu, Yixin Qin, Qian Xu, Shuai He, Bohong Cen, Wenjie Liao, Aimin Ji
ABSTRACT
In recent years, long noncoding RNAs (lncRNAs) have been shown to have critical roles in a broad range of cell biological processes. However, the activities of lncRNAs during ischemic stroke remain largely unknown. In this study, we carried out a genome-wide lncRNA microarray analysis in rat brains with ischemia/reperfusion (I/R) injury. The results revealed the differential expression of a subset of lncRNAs. Through the construction of lncRNA-mRNA co-expression networks, we identified lncRNA-N1LR as a novel I/R-induced lncRNA. The functions of lncRNA-N1LR were assessed by silencing and overexpressing this lncRNA in vitro and in vivo. We found that lncRNA-N1LR enhanced cell cycle progression and cell proliferation, and inhibited apoptosis in N2a cells subjected to in vitro ischemia (oxygen-glucose deprivation/reoxygenation, OGD/R). Furthermore, we showed that lncRNA-N1LR reduced neuronal apoptosis and neural cell loss in I/R-induced mouse brains. Mechanistically, we discovered that lncRNA-N1LR promoted neuroprotection probably through the inhibition of p53 phosphorylation on serine 15 in a manner that was independent of its location-associated gene Nck1. In summary, our results indicated that lncRNA-N1LR promoted neuroprotection against ischemic stroke probably by inactivating p53. Thus, we propose that lncRNA-N1LR may serve as a potential target for therapeutic intervention following ischemic brain injury.
Sigma-Aldrich
MISSION® esiRNA, targeting human CHN1