MilliporeSigma
A Deep Insight into the Photoluminescence Properties of Schiff Base Cd
Inorganic chemistry (2017-10-24)
Ishani Majumder, Prateeti Chakraborty, Sanchari Dasgupta, Chiara Massera, Daniel Escudero, Debasis Das
ABSTRACT
A tridentate N,N,O donor ligand 2,4-dichloro-2-[(2-piperazine-4-yl-ethylimino)-methyl]-phenol (HL) was designed, and eight new Zn
Sigma-Aldrich
3,5-Dichlorosalicylaldehyde, 99%