MilliporeSigma
  • Organometallic nanoprobe to enhance optical response on the polycyclic aromatic hydrocarbon benzo[a]pyrene immunoassay using SERS technology.
Organometallic nanoprobe to enhance optical response on the polycyclic aromatic hydrocarbon benzo[a]pyrene immunoassay using SERS technology.
Environmental science and pollution research international (2014-08-12)
Mohamed Dribek, Emmanuel Rinnert, Florent Colas, Marie-Pierre Crassous, Néné Thioune, Catalina David, Marc de la Chapelle, Chantal Compère
ABSTRACT
We demonstrated the use of a new organometallic nanoprobe for competitive surface-enhanced Raman scattering (SERS) immunoassay devoted to the detection of polycyclic aromatic hydrocarbons (PAH) such as benzo[a]pyrene (BaP) in seawater. The nanoprobes are gold nanoparticles (GNPs) labeled by a Raman reporter, the 5,5'-dithiobis(succinimidyl-2-nitrobenzoate) (DSNB) and functionalized with monoclonal antibodies anti-BaP. The antibodies are bound with a high specificity to the analyte while the GNPs enhanced the Raman scattering of the DSNB. This type of immunoassay involved the grafting of BaP onto a sensing surface. Thus, NH
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
N-Methylformanilide, 99%
Sigma-Aldrich
DCC, 99%