Merck
Matrix stiffness regulates migration of human lung fibroblasts.
Physiological reports (2017-05-17)
Shuichi Asano, Satoru Ito, Kota Takahashi, Kishio Furuya, Masashi Kondo, Masahiro Sokabe, Yoshinori Hasegawa
ABSTRACT
In patients with pulmonary diseases such as idiopathic pulmonary fibrosis and severe acute respiratory distress syndrome, progressive pulmonary fibrosis is caused by dysregulated wound healing via activation of fibroblasts after lung inflammation or severe damage. Migration of fibroblasts toward the fibrotic lesions plays an important role in pulmonary fibrosis. Fibrotic tissue in the lung is much stiffer than normal lung tissue. Emerging evidence supports the hypothesis that the stiffness of the matrix is not only a consequence of fibrosis, but also can induce fibroblast activation. Nevertheless, the effects of substrate rigidity on migration of lung fibroblasts have not been fully elucidated. We evaluated the effects of substrate stiffness on the morphology,
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-phospho-Myosin Light Chain (pSer19) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution