Merck
  • LncRNA CASC2 Interacts With miR-181a to Modulate Glioma Growth and Resistance to TMZ Through PTEN Pathway.
LncRNA CASC2 Interacts With miR-181a to Modulate Glioma Growth and Resistance to TMZ Through PTEN Pathway.
Journal of cellular biochemistry (2017-01-26)
Yiwei Liao, Liangfang Shen, Haiting Zhao, Qing Liu, Jun Fu, Yong Guo, Renjun Peng, Lei Cheng
ABSTRACT
Temozolomide (TMZ)-based chemotherapy is a standard strategy for glioma, while chemoresistance remains a major therapeutic challenge. Recent evidence highlights the crucial regulatory roles of long non-coding RNAs (lncRNA) in tumor biology. However, the roles and regulatory mechanisms of lncRNA cancer susceptibility candidate 2 (CASC2), in glioma tumorigenesis and chemoresistance are poorly understood. In this study, CASC2 expression was down-regulated in glioma tissues and cell lines, and was related to a clinicopathologic features and shorter survival time. Exogenous CACS2 alone was sufficient to inhibit glioma cells' proliferation and amplified TMZ-induced repression of cell proliferation, while CACS2 knockdown could reverse this process. CACS2 overexpression could sensitize TMZ-resistant glioma cells to TMZ, while CACS2 knockdown exerted the opposite function. Moreover, CASC2 could inhibit the miR-181a expression by direct targeting in TMZ-resistant glioma cells. CASC2 up-regulated PTEN protein and down-regulated p-AKT protein through regulating miR-181a, and the effect of CASC2 on PTEN and p-AKT could be partially restored by miR-181a. With TMZ-resistant glioma tissues, miR-181a was up-regulated while PTEN was down-regulated. Taken together, these observations suggest CASC2 up-regulates PTEN through direct inhibiting miR-181a and plays an important role in glioma sensitivity to TMZ and may serve as a potential target for cancer diagnosis and treatment. J. Cell. Biochem. 118: 1889-1899, 2017. © 2017 Wiley Periodicals, Inc.
Sigma-Aldrich
Imprint® RNA Immunoprecipitation Kit, High-capacity Protein A magnetic beads for successful RNA Immunoprecipitation,suitable for use with mRNA and microRNA