MilliporeSigma
  • Estrogenic and anti-androgenic effects of the herbicide tebuthiuron in male Nile tilapia (Oreochromis niloticus).
Estrogenic and anti-androgenic effects of the herbicide tebuthiuron in male Nile tilapia (Oreochromis niloticus).
Aquatic toxicology (Amsterdam, Netherlands) (2017-11-24)
Milena Devechi de Almeida, Thiago Scremin Boscolo Pereira, Sergio Ricardo Batlouni, Camila Nomura Pereira Boscolo, Eduardo Alves de Almeida
ABSTRACT
Tebuthiuron is a phenylurea herbicide widely used in agriculture that can reach the aquatic environments, possibly posing negative effects to the aquatic biota. Phenylurea herbicides, such as diuron, are known to cause estrogenic and anti-androgenic effects in fish, but no such effects were yet reported for tebuthiuron exposure. Thus, the aim of this study was to evaluate if tebuthiuron, at environmentally relevant concentrations (100 and 200ng/L) and after 25days of exposure have estrogenic and/or anti-androgenic effects on male of Nile tilapia (Oreochromis niloticus), through the evaluation of plasmatic testosterone (T) and estradiol (E
Product Description
Supelco
Tebuthiuron, PESTANAL®, analytical standard