MilliporeSigma
  • ADAM12 induces EMT and promotes cell migration, invasion and proliferation in pituitary adenomas via EGFR/ERK signaling pathway.
ADAM12 induces EMT and promotes cell migration, invasion and proliferation in pituitary adenomas via EGFR/ERK signaling pathway.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2017-11-16)
Junwen Wang, Zhuo Zhang, Ran Li, Feng Mao, Wei Sun, Juan Chen, Huaqiu Zhang, Jörg-W Bartsch, Kai Shu, Ting Lei
ABSTRACT
Pituitary adenomas are the second most common primary brain tumor with invasive properties. We have previously identified that ADAM12 (a disintegrin and metalloprotease 12) overexpression is associated with the tumor invasion of pituitary adenomas, however, the underlying mechanism remains unknown. This study aims to elucidate the mechanistic role of ADAM12 in regulating the tumor invasion of pituitary adenomas. In this study, we first showed that ADAM12 expression was concomitant with epithelial to mesenchymal transition (EMT) process in clinical specimens of human pituitary adenomas. Further functional studies showed that ADAM12 silencing in pituitary adenoma cells significantly inhibited the EMT process and suppressed cell migration, invasion and proliferation without influencing cell apoptosis. Mechanistically, ADAM12 silencing significantly reduced ectodomain shedding of epidermal growth factor receptor (EGFR) ligands and attenuated the EGFR/ERK signaling pathway. Blocking of EGFR signaling resulted in EMT suppression similar to silencing of ADAM12 and reduced cell migration, invasion and proliferation, while EGFR activation abolished the suppression on EMT, proliferation, migration and invasion induced by ADAM12 silencing. Moreover, ADAM12 silencing significantly impaired tumorigenesis and EMT of pituitary adenoma cells in vivo. Taken together, our study provide crucial evidence that ADAM12 induces EMT and promotes cell migration, invasion and proliferation in pituitary adenomas via EGFR/ERK signaling pathway. These finds strongly suggest that ADAM12 might serve as a novel valuable therapeutic target for pituitary adenomas.
Sigma-Aldrich
MISSION® esiRNA, targeting human ADAM12