MilliporeSigma
  • Testosterone-like immunoreactivity in hair measured in minute sample amounts - a competitive radioimmunoassay with an adequate limit of detection.
Testosterone-like immunoreactivity in hair measured in minute sample amounts - a competitive radioimmunoassay with an adequate limit of detection.
Scientific reports (2017-12-17)
Julia K Slezak, Jakob O Ström, Elvar Theodorsson
ABSTRACT
The concentrations of testosterone deposited in hair during hair growth may provide a retrospective reflection of the concentrations of bioactive testosterone in plasma. The objective of this study was to develop a radioimmunoassay with a sufficiently low limit of detection to measure the testosterone-like immunoreactivity in smaller hair samples (5 mg) than used in earlier studies, and to compare three different extraction procedures. The competitive radioimmunoassay consisted of a polyclonal antiserum (immunogen testosterone-7α-BSA) and a radioligand synthesised from testosterone-3-CMO-histamine. The within-assay and total coefficients of variation in the working range was 3% and 4.5%, respectively. The limit of detection was 0.87 pg/mL, which is equivalent to 0.12 pg/mg testosterone in 5 mg of hair. The concentration of testosterone-like immunoreactivity in hair samples was 1.23 (SD 0.47) pg/mg in women and 2.67 (SD 0.58) pg/mg in men (pulverised hair). Significantly improved precision was found when pulverised hair was used compared to non-pulverised hair. Our data indicate that pulverisation of the hair prior to hormone extraction is crucial. Detection limits fit for the intended purpose are achievable with 5 mg samples of hair.
Product Description
Supelco
Testosterone solution, 1.0 mg/mL in 1,2-dimethoxyethane, drug standard