MilliporeSigma
  • Cholecystokinin Expression in the Development of Postinfarction Heart Failure.
Cholecystokinin Expression in the Development of Postinfarction Heart Failure.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2017-11-14)
Xiaoying Dong, Can Wang, Jingqi Zhang, Siyue Wang, Hongjian Li, Yang Kang, Shuangquan Tian, Lu Fu
ABSTRACT
Cholecystokinin (CCK) is expressed in cardiomyocytes and may also play an important role in cardiovascular regulation. Clinical studies have shown that plasma CCK levels are an independent marker of cardiovascular mortality in cardiac disease. However, whether the development of postinfarction heart failure is associated with changes in CCK expression is unknown. To investigate CCK expression patterns and the association between CCK expression and heart functional parameters, we randomized male Sprague-Dawley rats into myocardial infarction (MI) or sham operation (SO) groups. CCK expression levels were assessed by western blotting, immunohistochemistry, real-time polymerase chain reaction (PCR), and enzyme-linked immunosorbent assay (ELISA) at different time points (2, 4 or 6 weeks) after surgery. Brain natriuretic peptide (BNP) concentrations were determined using Western blotting and ELISA, myocardial morphology was assessed by microscopy. Plasma CCK and BNP levels were significantly increased in all the MI groups compared with the corresponding SO groups. However, the degree to which myocardial CCK mRNA and protein expression levels were increased the MI groups compared with the SO groups was greater in the 4- and 6-week groups than in the 2-week group. Furthermore, plasma CCK levels were positively correlated with BNP concentrations and left ventricular end-systolic diameter (LVDs) and significantly negatively correlated with the ejection fraction (EF) and shortening fraction (SF) in model animals. Heart failure progression after infarction is associated with upregulated CCK levels; thus, CCK may be useful as a novel marker of heart failure.
Sigma-Aldrich
(+)-Biotin 4-nitrophenyl ester, 98%