MilliporeSigma
  • A potentiometric immunosensor for enterovirus 71 based on bis-MPA-COOH dendrimer-doped AgCl nanospheres with a silver ion-selective electrode.
A potentiometric immunosensor for enterovirus 71 based on bis-MPA-COOH dendrimer-doped AgCl nanospheres with a silver ion-selective electrode.
The Analyst (2017-12-12)
Ai-Li Sun
ABSTRACT
Herein a new potentiometric immunoassay for the point-of-care detection of enterovirus 71 (EV71) was developed by using a silver (Ag
Sigma-Aldrich
2-Methylheptane, 98%
Sigma-Aldrich
2,2-Bis(hydroxymethyl)propionic acid, 98%