Merck
  • [Determination of mannitol in health food by capillary gas chromatography].
[Determination of mannitol in health food by capillary gas chromatography].
Sichuan da xue xue bao. Yi xue ban = Journal of Sichuan University. Medical science edition (2006-06-10)
He-xing Wang, Yuan-qian Li, Lei Li
ABSTRACT
To develop a capillary gas chromatographic method for the determination of mannitol in health food. Mannitol was extracted from the sample with water by ultrasonic wave. The aliquot of sample solution was acetylated to form mannitol hexacetate with lower boiling point. Then the derivative was separated with the use of SPB-608 capillary chromatographic column and determined by flame ionization detector (FID) at the column temperature of 180 degrees C, the detector temperature of 210 degrees C and the nitrogen flow rate of 30 mL/min. The linear range of this method was 0.05-10.00 mg/mL and the limit of detection of was 0.01 mg/mL. The within-day and between-days precisions were 4.4% and 7.3%-7.6%, respectively. The average recoveries were 84.6%-107.6%. Mannitol could be separated from glucose, inositol and sorbitol under the optimum chromatographic condition. The results demonstrate that the method has the advantages of simplicity, accuracy and sensitivity. It is suitable for the determination of mannitol in health food.
Product Description
Supelco
SPB®-608 Capillary GC Column, L × I.D. 30 m × 0.53 mm, df 0.50
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPB®-608 Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.25 μm