MilliporeSigma
  • Enantiomeric separation in comprehensive two-dimensional gas chromatography with accurate mass analysis.
Enantiomeric separation in comprehensive two-dimensional gas chromatography with accurate mass analysis.
Chirality (2014-01-15)
Sung-Tong Chin, Yada Nolvachai, Philip J Marriott
ABSTRACT
Chiral comprehensive two-dimensional gas chromatography (eGC×GC) coupled to quadrupole-accurate mass time-of-flight mass spectrometry (QTOFMS) was evaluated for its capability to report the chiral composition of several monoterpenes, namely, α-pinene, β-pinene, and limonene in cardamom oil. Enantiomers in a standard mixture were fully resolved by direct enantiomeric-GC analysis with a 2,3-di-O-methyl-6-t-butylsilyl derivatized β-cyclodextrin phase; however, the (+)-(R)-limonene enantiomer in cardamom oil was overlapped with other background components including cymene and cineole. Verification of (+)-(R)-limonene components based on characteristic ions at m/z 136, 121, and 107 acquired by chiral single-dimension GC-QTOFMS in the alternate MS/MSMS mode of operation was unsuccessful due to similar parent/daughter ions generated by interfering or co-eluting cymene and cineole. Column phases SUPELCOWAX, SLB-IL111, HP-88, and SLB-IL59, were incorporated as the second dimension column ((2)D) in chiral GC×GC analysis; the SLB-IL59 offered the best resolution for the tested monoterpene enantiomers from the matrix background. Enantiomeric ratios for α-pinene, β-pinene, and limonene were determined to be 1.325, 2.703, and 1.040, respectively, in the cardamom oil sample based on relative peak area data.
Product Description
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 15 m × 0.53 mm, df 1.00 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 60 m × 0.25 mm, df 0.25 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 0.25 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 60 m × 0.53 mm, df 2.00 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 15 m × 0.32 mm, df 0.25 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 0.50 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 1.00 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.25 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 30 m × 0.53 mm, df 2.00 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 15 m × 0.10 mm, df 0.10 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 30 m × 0.25 mm, df 0.50 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 0.50 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 30 m × 0.53 mm, df 1.00 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 60 m × 0.53 mm, df 1.00 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 15 m × 0.25 mm, df 0.25 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 30 m × 0.20 mm, df 0.20 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 0.25 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 30 m × 0.53 mm, df 0.50 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 1.00 μm
Supelco
SUPELCOWAX 10 Capillary GC Column, L × I.D. 100 m × 0.25 mm, df 0.25 μm
Supelco
SLB®-1ms Capillary GC Column, L × I.D. 15 m × 0.32 mm, df 0.25 μm
Supelco
SLB®-1ms Capillary GC Column, L × I.D. 30 m × 0.32 mm, df 0.10 μm
Supelco
SLB®-1ms Capillary GC Column, L × I.D. 60 m × 0.32 mm, df 0.25 μm