MilliporeSigma
  • Development of a gas chromatography-mass spectrometry method for the determination of pesticides in gaseous and particulate phases in the atmosphere.
Development of a gas chromatography-mass spectrometry method for the determination of pesticides in gaseous and particulate phases in the atmosphere.
Analytica chimica acta (2011-06-28)
E Borrás, P Sánchez, A Muñoz, L A Tortajada-Genaro
ABSTRACT
A reliable multi-residue method for determining gaseous and particulate phase pesticides in atmospheric samples has been developed. This method, based on full scan gas chromatography-mass spectrometry (GC-MS), allowed the proper determination of sixteen relevant pesticides, in a wide range of concentrations and without the influence of interferences. The pesticides were benfluralin, bitertanol, buprofezin, chlorfenvinphos, chlorpyrifos, chlorpyrifos-methyl, ethalfluralin, fenthion, lindane, malathion, methidathion, propachlor, propanil, pyriproxifen, tebuconazol and trifluralin. Comparisons of two types of sampling filters (quartz and glass fibre) and four types of solid-phase cartridges (XAD-2, XAD-4, Florisil and Orbo-49P) showed that the most suitable supports were glass fibre filter for particulate pesticides and XAD-2 and XAD-4 cartridges for gaseous pesticides (>95% recovery). Evaluations of elution solvents for ultrasonic-assisted extraction demonstrated that isooctane is better than ethylacetate, dichloromethane, methanol or a mixture of acetone:hexane (1:1). Recovery assays and the standard addition method were performed to validate the proposed methodology. Moreover, large simulator chamber experiments allowed the best study of the gas-particle partitioning of pesticides for testing the sampling efficiency for the validation of an analytical multiresidue method for pesticides in air. Satisfactory analytical parameters were obtained, with a repeatability of 5±1%, a reproducibility of 13±3% and detection limits of 0.05-0.18 pg m(-3) for the particulate phase and 26-88 pg m(-3) for the gaseous phase. Finally, the methodology was successfully applied to rural and agricultural samples in the Mediterranean area.
Product Description
Supelco
ORBO 609 Amberlite XAD®-2 (20/50) 400/200 mg, W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
Supelco
ORBO 80 Coated Filter, with cassettes unassembled
Supelco
ORBO 402 Tenax® TA specially treated (35/60), 100/50 mg, W,W,W separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 49P (OVS) Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 270/140 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS) Tube, pkg of 10 ea
Supelco
ORBO 554 H2SO4 on Silica Gel (20/40) 150/75 mg, W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 53 Activated Silica Gel (20/40) specially cleaned, 400/200 mg with Glass Fiber Filters, GFF,F,F separators, O.D. × L 7 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 502 Activated Silica Gel (20/45) 100/50 mg, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 23 2-HMP on Amberlite XAD®-2 (20/40), 120/60 mg, W,W,W separators, O.D. × L 6 mm × 85 mm, pkg of 25 ea
Supelco
ORBO 43 Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 65P (OVS) Amberlite XAD®-4 specially cleaned, 160/80 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS), pkg of 10 ea
Supelco
ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
Supelco
ORBO 24 2-HMP on Amberlite XAD®-2 (20/40), 150/75 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 25 ea
Supelco
ORBO 80 Isocyanate Filter, with cassettes: no, filter O.D. 37 mm, pkg of 25 ea
Supelco
ORBO 52 Small Activated Silica Gel (20/40) 150/75 mg, pk 50, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 92 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group Vinyl acetate