MilliporeSigma
Microwave/SPME method to quantify pesticide residues in tomato fruits.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (2010-02-26)
Valérie Guillet, Céline Fave, Michel Montury
ABSTRACT
A new analytical method using focused microwave-assisted extraction (FMAE), coupled with solid phase micro-extraction (SPME), has been elaborated to determine 25 pesticides used in tomato cultivation. Microwave energy was used for a fast and controlled heating of solvent to selectively extract compounds. Calibration curves were plotted from blank tomato samples spiked at different concentrations with standards. A linear response was obtained between 10 and 1000 microg/Kg for pyrethroids and between 0.1 and 5000 microg/Kg for other compounds. For all studied substances, the resulting correlation coefficient (r(2)) was greater than 0.99. Limits of detection (LOD) and quantification (LOQ) were measured lower than 8 and 25 microg/Kg, respectively. The relative standard deviation (RSD) was determined below 15% for all pesticides. Field incurred tomato samples were used to validate the new FMAE/SPME method. Observed analysis results by using this technique were in good agreement compared to those obtained by two accredited trading laboratories using traditional methods. Four tomato samples, bought in a local market, were also tested with the FMAE/SPME method.
Product Description
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, StableFlex, for use with autosampler
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME Sampling Stand, for use with 4 mL vials
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with autosampler
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with manual holder
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 23 ga
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with autosampler
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 30 μm(PDMS, needle size 24 ga, for use with autosampler
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), fused silica fiber, df 65 μm(PDMS/DVB, for use with manual holder, needle size 23 ga
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, for use with manual holder
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with manual holder
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, StableFlex, for use with manual holder
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, StableFlex, for use with autosampler
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME Sampling Stand, for use with 15 mL vials
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, StableFlex, for use with autosampler
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 24 ga, for use with manual holder
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with manual holder, needle size 23 ga
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, for use with autosampler, needle size 23 ga
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 23 ga
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 23 ga, StableFlex, for use with autosampler
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with autosampler
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 23 ga, PDMS/DVB StableFlex, for use with autosampler
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Divinylbenzene/Carboxen/Polydimethylsiloxane (DVB/CAR/PDMS), needle size 23 ga, StableFlex, for use with manual holder or autosampler, fiber L 2 cm
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 23 ga, for use with autosampler
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, needle size 24 ga, for use with manual holder
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 24 ga, for use with autosampler
SKU
Pack Size
Availability
Price
Quantity
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, for use with manual holder, StableFlex fiber