MilliporeSigma
  • Expansion of murine T cells bearing a unique T cell receptor beta-chain in Friend virus-induced tumor in situ.
Expansion of murine T cells bearing a unique T cell receptor beta-chain in Friend virus-induced tumor in situ.
Journal of immunology (Baltimore, Md. : 1950) (1992-05-01)
M Suzuki, H Koseki, Y Mizutani, K Kuribayashi, M Kanno, M Taniguchi
ABSTRACT
Heterogeneity of V alpha 1+ and V beta 10+ TCR alpha beta-chains, which are predominantly used in anti-FBL-3 CTL clones established in vitro, was investigated at a nucleotide level in FBL-3 tumor-infiltrating lymphocytes (TIL) in vivo. The majority (90%) of V beta 10+ beta-chains dominated in TIL used homogeneous V beta 10D beta 2.1 sequences identical to that used in the T cell clones with cytotoxic functions. The homogeneous TCR beta-chain expression was dominant and found to be about 10% of the total TCR beta-chains in the TIL population, which was a greater than 300-to 900-fold increase than in the regional lymph nodes. This is in good agreement with the in vitro data showing that about 11% CTL clones used the homogeneous V beta 10D beta 2.1+ beta-chain. However, the J beta segment does not seem to contribute greatly to the recognition and selection of this TCR because some of homogeneous VD+ beta-chains were associated with J beta segments other than J beta 2.7 of the CTL clones. The frequency of the V alpha 1J alpha 112-2+ alpha-chain expression of the CTL type was much less (3- to 80-fold increase compared to that of lymph node) and also varied in sample materials, indicating the lower contribution of the alpha-chain for the oligoclonality of the TCR. The results were also confirmed by quantitative PCR and RNase protection assays. This suggests that the dominant expression of the homogeneous TCR beta-chain is due to the expansion of the particular anti-FBL-3 CTL in the tumor in situ. Also, the TCR beta-chain, especially the V beta D beta region, rather than alpha-chain is more important for the recognition and selection of the anti-FBL-3 TIL with cytotoxic functions.