- Assignment of the human pancreatic colipase gene to chromosome 6p21.1 to pter.
Assignment of the human pancreatic colipase gene to chromosome 6p21.1 to pter.
Pancreatic colipase is a 12-kDa polypeptide cofactor for pancreatic lipase (EC 3.1.1.3), an enzyme essential for the absorption of dietary long-chain triglyceride fatty acids. Colipase is thought to anchor lipase noncovalently to the surface of lipid micelles, counteracting the destabilizing influence of intestinal bile salts. Using primers derived from the known amino acid sequence, we have used the polymerase chain reaction to produce a cDNA clone corresponding to the complete coding region of the human procolipase mRNA. Southern blot analysis of genomic DNA from a panel of mouse-human somatic cell hybrids indicated that the colipase gene (CLPS) resides on human chromosome 6. Further analysis of somatic cell hybrids carrying chromosome 6 translocations permitted regional localization of CLPS to the 6p21.1-pter region.