- Immunohistochemical analysis of cyclin A, cyclin D1 and P53 in mammary tumors, squamous cell carcinomas and basal cell tumors of dogs and cats.
Immunohistochemical analysis of cyclin A, cyclin D1 and P53 in mammary tumors, squamous cell carcinomas and basal cell tumors of dogs and cats.
The involvement of cyclin A, cyclin D1 and p53 proteins in canine and feline tumorigenesis was analyzed immunohistochemically. In the present study, a total of 176 cases were examined, among which there were 108 canine cases (75 mammary lesions, 16 squamous cell carcinomas and 17 basal cell tumors) and 68 feline cases (43 mammary lesions, 20 squamous cell carcinomas and 5 basal cell tumors). Speckled nuclear staining for cyclin A was observed in 19/38 (50%) canine malignant mammary tumors and 18/37 (48.6%) feline mammary carcinomas, while this was not seen in benign mammary tumors of either dogs or cats. Marked intense nuclear cyclin A staining was seen in 7/16 (43.8%) canine squamous cell carcinomas and 18/20 (90.0%) feline squamous cell carcinomas. Only 3/17 (17.6%) canine basal cell tumors showed slight and scattered staining for cyclin A. Expression of cyclin D1 was very rare in both canine and feline tumors. Nuclear staining of p53 was found in 7/37 (18.9%) feline mammary carcinomas. Intense immunoreactivity for p53 was found in 6/16 (37.5%) canine squamous cell carcinomas and 8/20 (40%) feline squamous cell carcinomas. These results suggest that cyclin A may have a role in the proliferation of canine malignant mammary tumors, feline mammary carcinomas and squamous cell carcinomas of dogs and cats, and p53 may associate with the tumorigenesis of feline mammary carcinomas and squamous cell carcinomas of dogs and cats.