Key Documents
7220020
Globe Scientific
Globe Scientific Diamond® RealSeal™ Round PP Carboys with Spigot
Carboy, Round with Spigot and Handles, PP, White PP Screwcap, 20 Liter, Molded Graduations, Autoclavable-1EA
About This Item
PDF Icon
Shipped & Sold By Globe Scientific LLC
A Trusted Marketplace Seller
PDF Icon
Have a question?
feature
with cap
material
Bottle: polypropylene
General description
Diamond® RealSeal™ Plastic Carboys from Globe Scientific meet the large volume liquid storage and dispensing needs of the most demanding laboratory applications. Rugged and durable, Diamond RealSeal Carboys are the safe, shatterproof alternative to glass for mixing, storing, and dispensing liquids in volumes from five to fifty liters. The carboys are available in a wide range of materials and designs suitable for use with most media, reagents, and solutions used in laboratories across the world.
  • Spigot allows simple and accurate dispensing of media and solutions
  • Round design maximizes mixing efficiency
  • Integrated wide handles facilitate easy carrying and pouring, even when wearing gloves
  • Easy to read, molded graduations
  • PP Carboys are autoclavable at 121°C and have the best chemical resistance