Skip to Content
MilliporeSigma

HFER-SE260-10A-10

Hoefer

Hoefer Mini Vertical Protein Electrophoresis Systems

SE260 Mini Complete unit with 1.0mm accessories

Shipped & Sold By Hoefer Inc.

A Trusted Marketplace Seller

Synonym(s):

Gel Electrophoresis, vertical electrophoresis, mini gel electrophoresis tank, protein separation

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Marketplace Support
Need help or have questions about this product?
Contact Seller


manufacturer/tradename

Hoefer Inc.

General description

Combines the advantages of small format and efficient cooling for rapid screening of proteins and nucleic acids

Features:

  • Maximum glass plate size 10 x 10.5 cm
  • Flexible design, additionally able to accommodate 10 x 8 cm format with optional accessories
  • Supports a wide variety of precast gels
  • Efficient active cooling ensures sharp bands
  • Run up to two gels (maximum 30 samples) at one time under identical conditions
  • Quick and easy to assemble

Complete Units Include:

  • Buffer Chamber/Cooling Core
  • Safety Lid with High Voltage Leads
  • Glass Plates, Rectangular, 10 x 10.5 cm, 10 pcs
  • Alumina Plates, Notched, 10 x 10.5 cm, 5 pcs
  • Combs, 10 well, 2 pcs
  • T-Spacers, 4 pcs
  • Gel caster

Basic Unit Includes:

  • The same as complete unit without the combs, spacers, plates, and caster.

Specifications:

  • Glass Plate Size (W x H): 10 x 10.5 cm or 10 x 8 cm
  • Spacers: 0.75 mm, 1.0 mm, 1.5 mm
  • Maximum Power Settings: 500 V, 500 mA, 12 W
  • Unit Dimensions (W x H x D): 16.5 x 18 x 16 cm


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library