Skip to Content
MilliporeSigma

HFER-SE600-15-15

Hoefer

Hoefer Standard Vertical Protein Electrophoresis Systems

SE600 Standard Dual Cooled Vertical Protein Electrophoresis Unit (Complete Unit)

Shipped & Sold By Hoefer Inc.

A Trusted Marketplace Seller

Synonym(s):

Gel Electrophoresis system, standard vertical electrophoresis, gel electrophoresis tank, protein separation system

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Marketplace Support
Need help or have questions about this product?
Contact Seller


manufacturer/tradename

Hoefer Inc.

General description

Standard vertical format for 18 cm long gels

Features:

  • Maximum glass plate size 18 x 16 cm
  • Produce straight lanes and sharp, well defined bands
  • Run up to four gels (maximum 112 samples) at one time under identical conditions
  • Run gels at uniform temperature from 1 to 45°C
  • Accommodates denaturing and native polyacrylamide gels and 2-D electrophoresis
  • A wide array of accessories allows you to tailor gel configurations to your needs

Complete Unit Includes:

  • Buffer Chamber
  • Safety Lid w/High Voltage Leads
  • Heat Exchanger
  • Glass Plates 18 x 16 cm, 6 pcs
  • Combs, 15-well, 1.5 mm thick, 2 pcs
  • Spacers, 1.5 mm thick, 2 cm wide, 4 pcs
  • Dual Gel Caster

Basic Unit Includes:

  • The same as complete unit (SE600-15-1.5) without the combs and spacers.

Specifications:

  • Glass Plate Size (W x H): 18 x 16 cm or 18 x 8 cm
  • Spacers: 0.75 mm, 1.0 mm, 1.5 mm
  • Maximum Power Settings: 1000 V, 500 mA, 50 W
  • Maximum Temperature: 45°C
  • Indoor Use: 4 to 40°C
  • Humidity: Up to 80%
  • Unit Dimensions (W x H x D): 32 x 29 x 14 cm


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library