Skip to Content
MilliporeSigma

HFER-SUB15

Hoefer

Hoefer Midi Horizontal Electrophoresis Systems

SUB15 Standard Submarine Gel Electrophoresis Unit

Shipped & Sold By Hoefer Inc.

A Trusted Marketplace Seller

Synonym(s):

Electrophoresis System, Gel Electrophoresis system, horizontal gel tank, DNA electrophoresis

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Marketplace Support
Need help or have questions about this product?
Contact Seller


manufacturer/tradename

Hoefer Inc.

General description

Provides the most commonly used tray size and comb options

Features and Benefits

  • The market standard—a 15 x 15 cm gel tray, with comb options ranging from 1 to 30 samples
  • Multi-channel pipette compatible combs—with a maximum 30 sample throughput reduce gel loading time
  • Buffer recirculation ports—may be connected to a peristaltic pump for buffer recirculation during electrophoresis to maintain buffer pH and prevent ionic gradient formation
  • UV-transparent running tray—allows the user to image the gel without risk of damage due to handling
  • Side handles—for safe and easy transportation

Includes:

  • Buffer Chamber Assembly
  • Safety Lid
  • High Voltage Leads
  • Running Tray with Casting Gates
  • 16-well combs, 2 pcs
  • Colored Loading Strips
  • Buffer Recirculation Ports, 2 pcs

Specifications:

  • Gel Dimensions (W x L): 15 x 15 cm
  • Maximum Buffer Volume: 1.2 L
  • Maximum Power Settings: 300 V, 200 mA, 60 W
  • Maximum Sample Capacity: 120
  • Unit Dimensions (W x L x H): 21.5 x 33.5 x 7 cm


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library