Skip to Content
MilliporeSigma

HFER-TE22

Hoefer

Hoefer Wet Tank Transfer Sytems

TE22 Mighty Small Wet Transfer Tank

Shipped & Sold By Hoefer Inc.

A Trusted Marketplace Seller

Synonym(s):

Blotting tank, transfer system, western blotting, northern blotting, southern blotting

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Marketplace Support
Need help or have questions about this product?
Contact Seller


manufacturer/tradename

Hoefer Inc.

General description

Quickly and efficiently transfers proteins and nucleic acids from small gels to nylon, nitrocellulose or PVDF membranes

Features and Benefits:

  • Transfer as many as four small gels, up to 9 x 10 cm in less than an hour
  • Uniform and strong electric field—supports efficient and even transfers
  • Color-coded, easy to assemble and load cassettes—ensures proper orientation during transfer
  • Superior tank design allows the cassettes to apply equal pressure across the stack—prevents gel distortion
  • Built-in ceramic heat exchanger—provides excellent temperature control when used with an external cooling water bath
  • Built-in alumina-covered cooling channel provides excellent temperature control with no more than a 5°C temperature increase during a typical run

Includes:

  • Lower Chamber with Heat Exchanger
  • Safety Lid with High Voltage Leads
  • Electrode Panels, 2 pcs
  • Cassettes, 4 pcs
  • Foam Sponges, 6 mm thick, 4 pcs
  • Foam Sponges, 3 mm thick, 8 pcs
  • Cassette Hook
  • Blotter Paper (9 x 10.5 cm), 25 sheets


Still not finding the right product?

Explore all of our products under Hoefer Wet Tank Transfer Sytems




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library