MilliporeSigma
49101-07
MicroSolv
MicroSolv Chemical Solutions Brand Qualification and Calibration Kits
HSQ Kit, Complete. 75mm HPLC column, test solutions, accessories & software for performance qualification. Chemical Solutions brand. 1 EA.
Synonym(s):
PQ
Sign In to View Organizational & Contract Pricing.
Select a Size
Product Name
MicroSolv Chemical Solutions Brand Qualification and Calibration Kits, HSQ Kit, Complete. 75mm HPLC column, test solutions, accessories & software for performance qualification. Chemical Solutions brand. 1 EA.
General description
This complete high-speed qualification (HSQ™) kit for self-compliance of HPLC performance qualification (PQ) provides everything you need to complete a full qualification of your HPLC instruments and perform holistic and modular AIQ tests - in less than half a workday. You can do preventative maintenance with immediate instrument uptime without waiting for vendors to verify your instrument. Fulfill your requirements to “maintain instruments in a qualified state.” See USP <1058>. You can use this kit to qualify your HPLC Instruments for precision and accuracy.
The HSQ Kit includes:
  • 6 Linearity Solutions - 30 ml (L2 and L3 60 ml)
  • 1 Resolution Test Mixture - 30 ml
  • 1 Wavelength Calibration Solution (Holmium Oxide) - 4 ml
  • 1 Gradient Visualization Solution - 30 ml
  • 1 Cogent HPLC Certified Test Column - 75 mm x 4.6 mm
  • 1 Software Template. Excel with Operator Determined Criteria
  • 1 Accessories for Flow Cell Filling Pump Kit
  • 1 Quick Start Instructions
With proper use, you can perform approximately 25 qualifications per kit.
Solutions will be supplied with at least six months until the expiration date. They are designed to be used promptly and often. Expiration date is two years after the date of manufacture.
Choose from one of the most recent versions:
Certificates of Analysis (COA)