Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

70000-15P-2

MicroSolv

MicroSolv Cogent Diamond Hydride HPLC Columns

HPLC Column, Diamond Hydride, 4 um, 2.1 mm ID x 150 mm length, 100 A. Cogent TYPE-C Brand. 1 EA.

Synonym(s):

HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

PDF Icon
A Trusted Marketplace Seller
PDF Icon
Have a question?

technique(s)

HPLC

General description

This Cogent Diamond Hydride HPLC Column is made with Cogent TYPE-C™ Silica, and this silica hydride based stationary phase provides excellent Selectivity, Faster Equilibration and Robust Column Lifetimes.

Chosen for analysis of polar compounds that are difficult to retain in Reversed Phase, including polar and non-polar peptides, amino acids and other biological compounds such as metabolites. When using in Aqueous Normal Phase, this column is Mass Spec Friendly.

This column is often chosen for LCMS when increased resolution is desired.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library