Key Documents
19-042E
Omni International
Omni Bead Ruptor Elite 115-230VAC, 50/60Hz
About This Item
PDF Icon
Shipped & Sold By Omni International
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
Omni International
General description
Lyse the toughest samples in seconds with The Bead Ruptor Elite. The Bead Ruptor Elite is the most advanced and easy to use bead mill homogenizer available. It is specifically designed for grinding, lysing, and homogenizing biological samples prior to molecular extraction.
  • Wide range of accessories for any sample size - 24 x 0.5 mL, 24 x 1.5 mL, 24 or 48 x 2 mL, 12 x 7 mL, 12 x 15 mL, 6 x 30 mL, 3 x 50 mL.
  • Most powerful bead mill on the market – homogenize the most challenging samples
  • New touch screen interface. Sample Preparation at your fingertips.
  • Optimate™ sample prep software
  • No cool down required between runs: process hundreds of samples per day
  • Convenient front loading design, 99 programmable memory settings, and multilingual user interface
  • Features an integrated lid lock to prevent lid opening during operation
  • The ideal homogenizer for preparing samples when extracting DNA, RNA, proteins, and small molecules
  • Optimized tube motion ensures highest performance