Key Documents
TH115
Omni International
Omni 115 volt Tissue Homogenizer
About This Item
PDF Icon
Shipped & Sold By Omni International
A Trusted Marketplace Seller
PDF Icon
Have a question?
manufacturer/tradename
Omni International
General description
The redesigned Omni TH is a powerful Tissue Homogenizer designed to handle a wide range of laboratory samples from 0.2 to 100 mL in volume. The TH can usually process a tissue sample in less than 30 seconds and is compatible with a wide variety of generator probes for flexibility in application requirements. The Omni TH will accommodate a wide range of lab applications such as tissue homogenization, emulsions and suspensions, cell disruption, protein extraction, and medical research. It is lightweight enough to be hand-held and can also be mounted to a motor stand for lengthier processing.