OPT-AC2
Oxford Lab Products
Oxford Lab PCR Tubes
OPTimate 0.2 mL 8-Strip PCR tubes with Attached Optical Flat Caps
About This Item
Recommended Products
General description
Oxford Lab OptiMate PCR/qPCR Strip Tubes
OptiMate PCR Strip Tubes offer the design and security needed for high-performance PCR or qPCR work. OptiMate PCR Strip Tubes have the compatibility you need!
Features
- Convenient and Easy-to-Access packaging that includes organized tube strips.
- Ultra-thin walls allows sample to be efficiently and thoroughly heated, effectively and consistently yielding good results.
- OptiMate PCR/qPCR Strip Tubes are manufactured in highly clean areas and with highly clean plastic to avoid contaminants.
- Wall thickness is uniform throughout OptiMate PCR/qPCR Strip Tubes to enable consistent and repeatable results over multiple iterations of use.
- OptiMate PCR/qPCR Strip Tubes are available with attached caps, separate caps, or as tubes or caps purchased separately.
Properties
- Ultra-Thin and Uniform Walls enables optimal heat transfer between machine and sample
- Free of Contaminants DNase, RNase, Human DNA, PCR Inhibitors, and Endotoxins free
- Non-Reactive Internal Surface avoids accidental reactions
- Wide Compatibility Range multiple compatibility options for common thermocyclers
- Multiple Configurations multiple size and design options for compatibilit
OptiMate PCR/qPCR Strip Tubes are designed to be the reliable and high-performing consumables needed to optimize your PCR/qPCR procedures. Our PCR/qPCR Strip Tubes are designed with the specific needs of the user in mind – taking special care to maximize the yield of the thermocycler interaction. The plastic molding takes into account the need for ultra-thin walls to make sure that samples are effectively and efficiently heated and cooled. Likewise, the molds are finely refined to ensure that the thickness of the plastic is uniform at all points, so that samples are consistently heated and the yield remains repeatable across all tubes and across future uses.
Plastics must be low-risk and high confidence, so OptiMate PCR/qPCR Strip Tubes are carefully manufactured to ensure that contamination can be avoided. The quality process ensures that products avoid common contaminants. The plastic in OptiMate PCR/qPCR Strip Tubes is carefully chose and processed to further avoid any contamination. Finally, the packaging is carefully limited to small amounts of tubes so that users can open small amounts of tubes and avoid exposing their full inventory of tubes to possible contamination.