Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

EC-0100

Oxyrase

Oxyrase Enzyme System

1x100mL, Sterile 30u/ml

Sign Into View Organizational & Contract Pricing


About This Item

PDF Icon
Shipped & Sold By Oxyrase
A Trusted Marketplace Seller
PDF Icon
Have a question?

biological source

Escherichia coli

storage temp.

-20 °C

storage condition

avoid repeated freeze/thaw cycles

manufacturer/tradename

Oxyrase

General description

Oxyrase® is an enzyme system that offers a way to remove dissolved oxygen from liquid, gas or semisolid products. Oxyrase® is designed to work efficiently over a wide range of pH and temperature.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library