Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30-1130

TriAltus™

TriAltus™ SUMO Protease

1 mg, Yeast protein overexpressed in E. coli with histidine (His) tag

Synonym(s):

SUMO, SUMO Protease, SUMO P, Ulp, Ulp1, Enzyme, SUMO Enzyme, Protease

Sign Into View Organizational & Contract Pricing


About This Item

PDF Icon
Shipped & Sold By TriAltus Bioscience
A Trusted Marketplace Seller
PDF Icon
Have a question?

General description

SUMO Protease can be used for cleaving both natural SUMO-modified proteins and recombinant SUMO fusion proteins. This powerful site-specific protease targets the tertiary structure of the Ubiquitin-like SUMO domain. It hydrolyzes the peptide bond in the x–Gly–Gly–x sequence after the di-glycine bond, ensuring precise cleavage without leaving any residual amino acids.


SPECIFICATIONS

PURITY | ~100% (SDS gel analysis)

ENZYME SOURCE | Yeast protein overexpressed in E. coli with histidine (His) tag

CLEAVAGE SITE | SUMO Protease cleaves after the C-terminal glycine at the end of the SUMO sequence. If the target protein is fused downstream of the SUMO C-terminus, SUMO Protease cleavage renders the native protein sequence without any additional amino acids.

CLEAVAGE EFFICIENCY | 1 µg protease will digest 2 mg protein with 99% efficiency at 4 °C in 40 minutes.

STORAGE/SHIPPING CONCENTRATION | ~7.5 mg/mL

SHIPPING CONDITIONS | We ship SUMO protease on dry ice in buffer (15 mM Tris (pH 8.0), 0.2 M NaCl, 50% glycerol).

RECOMMENDED STORAGE CONDITIONS | -80°C long term or -20°C for 2-3 months in shipping buffer

EXPIRATION | 6 months from receipt when stored as directed

SKU | 

1 mg: 30-1130

5 mg: 30-1135

10 mg: 30-1140

50 mg: 30-1146

100 mg: 30-1150


Additional processing information: This highly efficient SUMO protease is compatible with TriAltus™ CLiM protein affinity purification technology.

Legal Information

Use of TriAltus products falls under our limited license agreement and is for non-profit research use only. For commercial use, please contact us for licensing.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library