Skip to Content
MilliporeSigma

TRAB-30-1130

TriAltus™

TriAltus™ SUMO Protease

1 mg, Yeast protein overexpressed in E. coli with histidine (His) tag

Shipped & Sold By TriAltus Bioscience

A Trusted Marketplace Seller

Synonym(s):

SUMO, SUMO Protease, SUMO P, Ulp, Ulp1, Enzyme, SUMO Enzyme, Protease

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Marketplace Support
Need help or have questions about this product?
Contact Seller


General description

SUMO Protease can be used for cleaving both natural SUMO-modified proteins and recombinant SUMO fusion proteins. This powerful site-specific protease targets the tertiary structure of the Ubiquitin-like SUMO domain. It hydrolyzes the peptide bond in the x–Gly–Gly–x sequence after the di-glycine bond, ensuring precise cleavage without leaving any residual amino acids.

SPECIFICATIONS

PURITY | ≥95% (SDS-PAGE)

ENZYME SOURCE | Yeast protein overexpressed in E. coli with histidine (His) tag

CLEAVAGE SITE | SUMO Protease cleaves after the C-terminal glycine at the end of the SUMO sequence. If the target protein is fused downstream of the SUMO C-terminus, SUMO Protease cleavage renders the native protein sequence without any additional amino acids.

CLEAVAGE EFFICIENCY | 1 µg protease will digest 2 mg protein with ≥90% efficiency at 4 °C in 60 minutes.

STORAGE/SHIPPING CONCENTRATION | ~7.5 mg/mL

SHIPPING CONDITIONS | We ship SUMO protease on dry ice in buffer (15 mM Tris (pH 8.0), 0.2 M NaCl, 50% glycerol).

RECOMMENDED STORAGE CONDITIONS | -80°C long term or -20°C for 2-3 months in shipping buffer

EXPIRATION | 6 months from receipt when stored as directed

SKU | 

1 mg: 30-1130

5 mg: 30-1135

10 mg: 30-1140

50 mg: 30-1146

100 mg: 30-1150

Additional processing information: This highly efficient SUMO protease is compatible with TriAltus™ CLiM protein affinity purification technology.



Still not finding the right product?

Explore all of our products under TriAltus™ SUMO Protease




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library