Skip to Content
MilliporeSigma

TRAB-30-2130

TriAltus™

TriAltus™ PreScission Protease

1 mg, Human Rhinovirus (HRV) 3C Protease overexpressed in E. coli with a glutathione S-transferase (GST) tag

Shipped & Sold By TriAltus Bioscience

A Trusted Marketplace Seller

Synonym(s):

PreScission, PreScission Protease, PSC, PSC P, Enzyme, Precission Enzyme, Protease

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

Marketplace Support
Need help or have questions about this product?
Contact Seller


General description

The PreScission protease (PSC) is a human rhinovirus (HRV) type 14 3C protease fused to glutathione S-transferase (GST). This protease recognizes and cleaves the site specific sequence LEVLFQ/GP (where “/” indicates the cleavage site).

SPECIFICATIONS

PURITY | ≥95% (SDS-PAGE)

ENZYME SOURCE | Human Rhinovirus (HRV) 3C Protease overexpressed in E. coli with a glutathione S-transferase (GST) tag

CLEAVAGE SITE | PSC recognizes the sequence LEVLFQGP and cleaves between the glutamine (Q) and glycine (G)

CLEAVAGE EFFICIENCY | 20 µg protease will digest 2 mg protein with ≥90% efficiency at 4°C in 60 minutes.

STORAGE/SHIPPING CONCENTRATION | ~5 mg/mL

SHIPPING CONDITION | We ship PreScission Protease on dry ice in buffer (20 mM Tris (pH 8.0), 0.1 M NaCl, 1 mM EDTA, 50% glycerol, 1 mM βMe).

RECOMMENDED STORAGE CONDITIONS | -80 °C long term or -20 °C for 2-3 months in shipping buffer

EXPIRATION | 6 months from receipt when stored as directed


SKU | 

1 mg: 30-2130

5 mg: 30-2135

10 mg: 30-2140

50 mg: 30-2146

100 mg: 30-2150

Additional processing information: This highly efficient PreScission protease is compatible with TriAltus′ CLiM protein affinity purification technology.



Still not finding the right product?

Explore all of our products under TriAltus™ PreScission Protease




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library