Key Documents
VTGNPFB12-MD
Valutek
Valutek Double Chlorinated Powder-Free 12'' Nitrile Gloves
Medium, Double Chlorinated Disposable Powder Free 3.94 mil Nitrile Gloves , Class 100-1,000 (ISO 5-6), Case of 1000,
Synonym(s):
Nitrile Gloves, Disposable Gloves, Cleanroom Gloves
About This Item
PDF Icon
Shipped & Sold By Valutek
A Trusted Marketplace Seller
PDF Icon
Have a question?
material
nitrile
size
M
manufacturer/tradename
Valutek
General description
VTGNPFB12 is Valutek's 12-inch powder-free ambidextrous cleanroom nitrile glove. It is made from 100% clean synthetic nitrile polymer and does not contain any rubber latex, fillers, or silicones.
It features a beaded cuff and a textured fingertip design. The specially formulated high-modulus nitrile offers unmatched operator comfort and dexterity.
Packaged in a cleanroom.
All Valutek gloves are thoroughly tested and produced in ISO-compliant facilities, subject to Valutek's inspection and rigorous process control measures to uphold Valutek's quality standards and product specifications.
Product Features:
  • Crafted from 100% clean synthetic nitrile polymer (Acrylonitrile Butadiene)
  • Free from accelerators and sulfur
  • Completely devoid of fillers, silicones, or plasticizers
  • Measures 12 inches (290 mm) in length with a beaded long cuff
  • Equipped with textured fingertips for improved grip
  • Powder-free and undergoes double chlorination, including a filtered water rinse
  • Compliant with ESD (Electrostatic Discharge) standards and compatible with acids and solvents
  • Critical environment compatible.
  • Part number and lot number traceable for 36 months from the manufacturing date.
Product Application:
Valutek classifies this product as "Microtek" in terms of cleanliness level, and recommends it to use for  cleanroom Class 100-1,000 (ISO 5-6) critical environment. 
These gloves are commonly employed in a wide range of applications, including semiconductor manufacturing, pharmaceutical processes, food handling, laboratory work, electronics, intricate parts handling, and maintenance and cleanup tasks.
Product Packaging:
  • 100 ea/bag, 10 bags/case, 1000 ea/case.
  • The packaging consists of an outer bag containing an inner bag with two stacks of 50 gloves each.
  • Packaged with the cuffs positioned at the bottom and the thumbs facing inward.
  • Vacuum-sealed, flat-packed, and come with a carton liner for added protection.
  • Priced and sold by the case.
Choose from one of the most recent versions:
Certificates of Analysis (COA)