MilliporeSigma
VLSC-F20110481
Velp Scientifica
Velp Scientific Overhead Stirrer
OHS 100 Advance
Shipped & Sold By Velp Scientific, Inc.
A Trusted Marketplace Seller
Synonym(s):
Digital stirrer, overhead stirrer, digital overhead stirrer
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
manufacturer/tradename
Velp Scientifica
General description
The VELP OHS 100 Advance Overhead Stirrer is a premium, digitally controlled instrument designed for demanding, medium‑ to high‑viscosity stirring tasks. It combines powerful torque, precise control, advanced smart features, and robust safety—all in a compact benchtop design.
  • Brushless motor with SpeedServo™ ensures consistent torque across the full speed range.
  • Speed range: 30–1,300rpm, digitally controlled for precision stirring.
  • Powerful stirring capability: 100Ncm maximum torque, handles up to 100L and viscosities of 70,000mPas.
  • SmartChuck™ enables tool‑free, one‑handed shaft changes; automatically locks if opened during operation for safety.
  • 3.5″ graphical TFT display provides real‑time data speed, torque, temperature, and stirring profile graphs.
  • Programmable timer, with support for ramps and interval settings for unattended workflows.
  • Includes a Pt100 temperature probe; the stirrer can auto-adjust speed or stop when temperature thresholds are exceeded.
  • Connectivity: USB and Wi‑Fi interfaces, with integration into the VELP Ermes cloud platform via ControllerSoft™ for remote monitoring, method control, and data export.
  • Durable IP54-rated sealed housing with safety features: overload, over-temperature/current alarms, vibration sensor, safe‑start knob, and lock key—all built for rugged use. Backed by a 3 year warranty.
Still not finding the right product?
Explore all of our products under Velp Scientific Overhead Stirrer
Choose from one of the most recent versions:
Certificates of Analysis (COA)