Key Documents
MF34G-5
WPI
WPI MicroFil
34 ga, 67mm
About This Item
PDF Icon
A Trusted Marketplace Seller
PDF Icon
Have a question?
General description
Our MicroFil fills micropipettes easily and reliably. Its long and fine tip allows you to start the filling very close to the pipette tip, eliminating both air bubble formation and clogging due to the washing down of dust particles. The transparent amber MicroFil needle is constructed from a combination of plastic and fused silica - no metal components are used. The MicroFil plastic needle may be stored for days with the filling solution inside without clogging. Sold in packages of 5.
Choose from one of the most recent versions:
Certificates of Analysis (COA)