NANOFIL
WPI
WPI Sub-Microliter Injection System
NanoFil 10 µL Syringe, Use with NanoFil injection needles 33-36g
All Photos(1)
About This Item
Recommended Products
General description
The NanoFil TM sub-microliter injection system boasts zero dead volume . Variability is eliminated with the NanoFil TM design: our interchangable needles advance directly into the syringe barrel, creating a 1:1 connection with the plunger— a truly gas-tight system built for low-volume sample control.
Our cross-compatible NanoFil TM needles are offered in sizes from 33-36g, blunt or beveled