MilliporeSigma
WPI-NF36BV
WPI
WPI NanoFil Needles
Beveled, 36G
A Trusted Marketplace Seller
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Marketplace Support
Need help or have questions about this product?
Contact Seller
manufacturer/tradename
WPI
General description
The replaceable needles used with the NanoFil are available with either blunt or beveled tips. The blunt tip is used for injection into soft tissue and when a uniform solution distribution is needed. The beveled style is used for applications that involve the penetration of a tough tissue.
Still not finding the right product?
Explore all of our products under WPI NanoFil Needles
Choose from one of the most recent versions:
Certificates of Analysis (COA)