HomeGas Chromatography (GC) GC Analysis of Sweet Orange Essential Oil on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 µm), Fast GC Analysis
GC Analysis of Sweet Orange Essential Oil on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 µm), Fast GC Analysis
Fast GC Analysis of Sweet Orange Essential Oil on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm)
Conditions | |
---|---|
column | SLB-5ms, 10 m x 0.10 mm I.D., 0.10 µm (28465-U) |
oven | 40 °C, 50 °C/min to 320 °C |
carrier gas | hydrogen, 81.5 cm/sec constant |
sample | sweet orange essential oil in hexane |
injection | 0.4 µL, 300:1 split |
inj. temp. | 320 °C |
detector | FID, 320 °C |
Description | |
---|---|
Categories | Analytical Chromatography, GC Applications, Essential Oils |
Featured Industry | Flavors and Fragrances |
Legal Information | SLB is a registered trademark of Sigma-Aldrich Co. LLC |
Other Notes | Complementary products: Syringes Vials Gas Purification and Gas Management Inlet Liners, Glass Wool, and Specialized Hand Tools Septa and Specialized Hand Tools Column Ferrules, Nuts, and Specialized Hand Tools |
suitability | application for GC |
Analyte
- (-)-Perilla aldehyde (perillaldehyde)
- (-)-trans-Caryophyllene
- (-)-α-Copaene
- (-)-β-Pinene
- (+)-Sabinene
- (+)-Terpinen-4-ol
- (+)-Valencene
- (+)-δ-Cadinene
- ®-(+)-Citronellal
- ®-(+)-Limonene
- 1-Octanol
- 3,7-Dimethyl-2,6-octadienyl acetate
- 3-Carene
- Bicyclogermacrene
- Camphene
- cis-Limonene oxide
- Decanal
- Geranial
- Geranyl acetate
- Germacrene D
- Lauric aldehyde
- Linalool
- Myrcene
- Neral
- Neryl acetate
- Nonanal
- Octanal
- p-Cymene
- Perilla alcohol
- Terpinolene
- trans-2-Decenal
- trans-Limonene oxide
- trans-β-Farnesene
- trans-β-Ocimene
- Undecanal
- α-Huµlene
- α-Phellandrene
- α-Pinene
- α-Terpinene
- α-Terpineol
- α-Thujene
- β-Copaene
- γ-Terpinene