MilliporeSigma
HomePhotometry & ReflectometryDetermination of o-Phosphate in Ground and Surface Water
Sensitive Photometric Determination of ortho-Phosphate in Groundwater and Surface Water, Analogous to EPA 365.2+3, APHA 4500-P E, and DIN EN ISO 6878
Abstract
This protocol describes the sensitive determination of ortho-phosphate in groundwater and surface water using photometric analysis with ammonium molybdate. The procedure, based on EPA 365.2+3 and DIN EN ISO 6878 standards, utilizes Spectroquant® test kits and UV/VIS spectrophotometers for accurate and efficient phosphate measurement. The method effectively detects low concentrations of PO4-P, making it suitable for environmental water testing and monitoring.
Section Overview:
Introduction
Phosphorus is an essential element for organisms and plants.1 In natural, uncontaminated waters, it occurs as organically bound phosphate, condensed phosphates or as orthophosphate - often referred to by its chemical formula PO4-P. The small quantity of phosphorus present in natural waters does not promote the growth of plants. However, a rise in the concentration of phosphorus results in the proliferation of algae, which leads to the eutrophication of the water body.2, 3
In the mid-20th century, the anthropogenic contamination of water bodies with phosphate has resulted in widespread eutrophication via fertilizers, wastewater, and washing detergents, among other things. Measures were taken to reduce phosphate concentrations through discontinuation of the use of phosphates in detergents and the precipitation of phosphates in wastewater treatment plants, which brought about a reduction in the phosphate environmental burden of approximately 75%.2, 4
A prime example is the case of Lake Constance (Bodensee, Germany). At the end of the 1970s, the concentration of PO4-P in the lake was 84 μg/L, whereas today this major water body exhibits PO4-P levels of just 5-6 μg/L.5, 6
Spectroquant® spectrophotometer Prove 600 plus. The device has a sleek, modern design with a rectangular body. It is green with a black top and features a touchscreen interface displaying measurement data, as well as a slot for sample insertion and the "M" logo.
Spectroquant® Spectrophotometer Prove 600 plus
Analysis of phosphate in surface waters
The analysis of such low concentrations of PO4-P is challenging. According to DIN EN ISO 10304-1, the lower working range of the ion chromatography method is 33 μg/L PO4-P (equivalent to 100 μg/L PO4-), which is considerably higher than the phosphate concentrations usually present in surface waters.7 Photometry permits significantly more sensitive measurements. According to DIN EN ISO 6878, the lower measurement limit for the photometric determination of phosphorus in water is 5 μg/L PO4-P, if no laborious extraction procedure is performed.8
Reagents, Instruments and Materials
Phosphate Test Kit/Reagents
For the measurement, one of the following Spectroquant® test kits is necessary:
  • Spectroquant® Phosphate Test (1.14848)
Instrument(s) & Devices
For the measurement, one of the following Spectroquant® photometers is necessary:
  • Spectroquant® UV/VIS Spectrophotometer Prove 600 Plus (1.73028)
Also, legacy systems
  • Spectroquant® Spectrophotometer Prove 600
Software for Data transfer
  • Optional Spectroquant® Prove Connect to LIMS software package (Y.11086) to transfer your data into an existing LIMS system.
Instrument Accessories
  • Rectangular cells 100 mm (1.74011)
Other Reagents and Accessories
  • Water for analysis (1.16754)
  • Phosphate standard solution Certipur® (1.19898)
  • Preparation vessel, flat-bottomed long tubes with screw caps (1.14901)
  • Plastic Syringes, 20 mL, Luer slip, polypropylene (XX1102012)
  • Syringe filter holder, Millex-HPF LCR Filter, 0.20 μm, PTFE, with glass fiber prefilter (SLLGM25NS)
  • Pipettes for 20 mL with plastic tips or 20-mL volumetric pipettes
Experimental Procedure
Sample Preparation
  • Filter the sample as swiftly as possible after sampling over the above-mentioned filter into a clean and dry vessel.
Analysis
  • Determine the PO4-P content with the above mentioned test kit.
  • For measurement in the 100-mm cell, both the sample volume as well as the quantities of reagents PO4-1 and PO4-2 must be quadrupled, i.e., using a sample volume of 20 mL, 20 drops of reagent PO4-1 and 4 level microspoons of reagent PO4-2.
  • The measurement is carried out against a blank, prepared from water for analysis, and the reagents are prepared in an analogous manner.
  • After measurement, the result can be read off of the instrument.
Results
As a means to gain expressive statements on the suitability of the Spectroquant® test kit for the determination of the phosphate concentration in surface waters, six samples were investigated for their PO4-P content using this test kit. For comparison, a reference analysis according to the DIN EN ISO 6878 method was also performed. A detection limit of 0.003 mg/L PO4-P for the DIN method was determined in accordance with DIN (Table 1).
See more applications for photometry at Protocols and Application Notes
Groundwater and surface water
Concentration [mg/L PO4-P]
Spectroquant® Phosphate Test 1.14848
DIN EN ISO 6878
Lake Starnberger See
< 0.0025
< 0.0030
Lake Wörthsee
< 0.0025
< 0.0030
Lake Altmühlsee
0.0126
0.0130
Lake Brombachsee
0.0025
0.0056
Lake Ammersee
< 0.0025
< 0.0030
Dam North Rhine-Westfalia
< 0.0025
< 0.0030
Table 1.Comparison of results of the Spectroquant® Phosphate Test with a photometric reference analysis according to DIN EN ISO 6878
The two methods show comparable results: in four of the six samples, the PO4-P concentration was below the measurement range. In only two of the six samples were the measured values within the range. In the sample from the Brombach Lake, the detected concentrations differed by 0.0031 mg/L PO4-P. In the sample from Altmühl Lake, the deviation between the Spectroquant® result and the DIN result was a mere 0.0004 mg/L PO4-P. The recovery rate with regard to the DIN EN ISO method was 97%.
In addition to the reference analysis procedure, samples were also measured using the standard addition method. In this investigation, the five lake water samples and one reservoir sample were spiked with four different concentrations of PO4-P and the recovery in each sample was determined. Table 2 shows the results of this investigation.
Groundwater and surface water
Addition [mg/L PO4-P]
Recovered concentration [mg/L PO4-P]
Recovery rate
Lake Starnberger See
0.0050
0.0055
110%
0.0100
0.0101
101%
0.0500
0.0516
103%
0.1000
0.1005
101%
Lake Wörthsee
0.0050
0.0059
119%
0.0100
0.0103
103%
0.0500
0.0525
105%
0.1000
0.1025
102%
Lake Altmühl
0.0050
0.0066
131%
0.0100
0.0103
103%
0.0500
0.0523
105%
0.1000
0.1021
102%
Lake Brombachsee
0.0050
0.0069
137%
0.0100
0.0108
108%
0.0500
0.0520
104%
0.1000
0.1027
103%
Lake Ammersee
0.0050
0.0061
123 %
0.0100
0.0106
106 %
0.0500
0.0524
105 %
0.1000
0.1027
103 %
Dam North Rhine-Westfalia
0.0050
0.0059
119 %
0.0100
0.0112
112 %
0.0500
0.0532
106 %
0.1000
0.1056
106 %
Table 2.Standard addition with the Spectroquant® Phosphate Test
The added standard concentrations could be recovered in all samples. The average recovery rate was 105% ± 5%.
Note:
If the accuracy of the method is to be increased further, we recommend carrying out an own calibration for a limited measuring range. We recommend the performance of the calibration according to the ISO 8446-1 or respectively the DIN 38402-51 method.
For this purpose, a user-defined method can be programmed on the Prove 600 Plus.
For details, see Prove Manual chapter 9.6 “User-defined Concentration Methods”.
By recording a 10-point calibration curve for the measuring range 0.0005 - 0.0250 mg/L PO4-P, the following performance characteristics could be achieved:
a line graph titled "Calibration curve for the measurement range 0.0025 – 0.0250 mg/L PO4-P." The x-axis represents the concentration of PO4-P in mg/L, marked with values ranging from 0.0000 to 0.0250 at intervals of 0.0050 (0.0000, 0.0050, 0.0100, 0.0150, 0.0200, 0.0250). The y-axis represents absorbance (labeled as Absorbance A), marked with values ranging from 0.000 to 0.140 at intervals of 0.020 (0.000, 0.020, 0.040, 0.060, 0.080, 0.100, 0.120, 0.140).  The graph features a series of yellow dots representing data points, each corresponding to a specific concentration of PO4-P and its associated absorbance value. A solid green line runs through these points, indicating the calibration curve, which shows a linear relationship between the concentration of PO4-P and absorbance. Two dashed green lines run parallel to the solid line, representing the confidence intervals or margin of error around the calibration curve.  The data points follow a straight, upward-sloping line, indicating that as the concentration of PO4-P increases, the absorbance also increases, demonstrating a strong linear correlation between the two variables within the specified measurement range.
Figure 1.10-point calibration curve for the measuring range 0.0005 - 0.0250 mg/L PO4-P
The method shows a good linearity. Compared with the pre-programmed method, the performance characteristics could be considerably improved.
Pre-programmed method
0.0025 – 0.5000 mg/L PO4-P
User calibration
0.0025 - 0.0250 mg/L PO4-P
Method standard deviation [mg/L]
+ 0.0029
+ 0.0001
Method coefficient of variation [%]
+ 1.2
+ 0.88
Confidence interval (P = 95 %) [mg/L]
+ 0.006
+ 0.0003
Table 3.Performance characteristics
Summary
The Spectroquant® Phosphate Test (1.14848) is a rapid, inexpensive, and precise alternative to the standard methods for the determination of orthophosphate in groundwater and surface waters. The results are comparable with those obtained using the DIN EN ISO 6878 standard method. By using the standard addition method, it was demonstrated that the test kit is suitable for the quantification of PO4-P in groundwater and surface waters.
See more applications for photometry at Protocols and Application Notes
Related Products
Loading
References
1.
Belitz H, Grosch W. 1982. Wasser.1-6. https://doi.org/10.1007/978-3-662-08306-2_1
2.
Gleisberg D. 1988. Phosphate und Umwelt. Chemie in nserer Zeit. 22(6):201-207. https://doi.org/10.1002/ciuz.19880220604
3.
1999. Standard Methods for the Examination of Water and Wastewater, 4500P – Phosphorus. 18. Washington, DC: American Public Health Association, American Water Works Association, Water Environment Federation.. [Internet].
4.
Metzner G. 2006. Phosphate aus Wasch- und Reinigungsmitteln im kommunalen Abwasser der Bundesrepublik Deutschland. [Internet].
5.
2013. Internationale Gewässerschutzkommission für den Bodensee, Gesunder Bodensee dank niedriger Phosphorwerte. IGKB press release. [Internet].
6.
Bodensee-Wasserversorgung, [cited 24 Aug 2016]. [Internet]. Available from: http://www.zvbwv.de/index.php?id=135
7.
DIN EN ISO 10304-1:2009-07, Wasserbeschaffenheit_- Bestimmung von gelösten Anionen mittels Flüssigkeits-Ionenchromatographie_- Teil_1: Bestimmung von Bromid, Chlorid, Fluorid, Nitrat, Nitrit, Phosphat und Sulfat (ISO_10304-1:2007); Deutsche Fassung EN_ISO_10304-1:2009. https://doi.org/10.31030/1518948
8.
DIN EN ISO 6878:2004-09, Wasserbeschaffenheit – Bestimmung von Phosphor – Photometrisches Verfahren mittels Ammoniummolybdat (ISO 6878:2004). [Internet].
9.
2016. Package leaflet Spectroquant® Phosphate Test. [Internet]. Available from: https://www.sigmaaldrich.com/catalog/product/mm/114848?lang=en&region=US