HPLC Analysis of Warfarin™ Anticoagulant on Ascentis® C18
Conditions | |
---|---|
Column | Ascentis C18, 15 cm x 4.6 mm I.D., 5 µm particles (581324-U) |
Column Temp. | 35 °C |
Mobile phase | [A] 20 mM phosphoric acid (pH 2.0, unadjusted): [B] methanol; (30:70, A:B) |
Flow rate | 1 mL/min |
Sample | 50 µg/mL each in mobile phase |
Injection | 10 µL |
Detector | UV, 280 nm |
Description | |
---|---|
Categories | Analytical Chromatography, Pharmaceutical Analysis, Anticoagulants |
Featured Industry | Clinical Pharmaceutical (small molecule) |
Legal Information | Ascentis is a registered trademark of Sigma-Aldrich Co. LLC, Warfarin is a trademark of Wisconsin Alumni Research Foundation |
Suitability | Application for HPLC |