Merck
HomeEnzyme Activity AssaysEnzymatic Assay of Amyloglucosidase (EC 3.2.1.3)
Enzymatic Assay of Amyloglucosidase
(EC 3.2.1.3)
Description
This procedure may be used for the determination of Amyloglucosidase activity using starch as the substrate. The Spectrophotometric Stop Rate Determination [Absorbance at 340 nm (A340), Light path = 1 cm] is based on the following reactions:
     Amyloglucosidase
Starch + water ––––––––––––––––> D-Glucose
  Hexokinase
D-Glucose + ATP ––––––––––> Glucose-6-Phosphate + ADP
        G-6-PDH
Glucose-6-Phosphate + β-NADP ––––––––––> 6-PG + β-NADPH
ADP – Adenosine Diphosphate
ATP – Adenosine Triphosphate
G-6-PDH – Glucose-6-Phosphate Dehydrogenase
β-NADP – β-Nicotinamide Adenine Dinucleotide Phosphate, Oxidized form
β-NADPH – β-Nicotinamide Adenine Dinucleotide Phosphate, Reduced form
6-PG – 6-Phospho-D-Gluconate
Unit Definition – One unit of Amyloglucosidase will liberate 1.0 mg of glucose from starch in 3 minutes at pH 4.5 at 55 °C.
Precautions
Please consult the Safety Data Sheet for information regarding hazards and safe handling practices.
Reagents and Equipment Required
Sodium acetate, trihydrate (S8625)
Starch from potato (S2004)
6.1 N Trichloroacetic acid solution ~100% (w/v) (T0699]
Glucose Assay Reagent (G3293)
Sodium bicarbonate (S8875)
Preparation Instructions
Use ultrapure water (≥18 MΩxcm resistivity at 25 °C) for the preparation of reagents.
Buffer (50 mM Sodium Acetate, pH 4.5 at 55 °C) – Prepare a 6.8 mg/mL solution in ultrapure water using Sodium acetate, trihydrate (S8625). Adjust to pH 4.5 at 55 °C with 1 M HCl.
Starch Solution [1% (w/v)] – Prepare a 10 mg/mL solution in Buffer using soluble Starch from potato (S2004). Facilitate solubilization by heating for ~15 minutes at 60–80 °C. Do not boil. Allow the solution to slowly mix throughout the assay.
Enzyme Solution (Amyloglucosidase) – Immediately before use, prepare a solution (0.40–0.80 mg solid/mL) in cold ultrapure water. Then immediately dilute to 0.3–0.6 units/mL of Amyloglucosidase in 55 °C purified water. The final reaction mixtures must contain 0.15–0.60 units of Amyloglucosidase.
TCA Solution [(50% (w/v) Trichloroacetic Acid Solution] – Prepare a 2-fold dilution of 6.1 N Trichloroacetic acid solution ~100% (w/v) (T0699] in ultrapure water.
Glucose Assay Reagent (HK) – Immediately before use, dissolve the contents of one vial of Glucose Assay Reagent (G3293) with ultrapure water using volume indicated by package size on label.
Procedure
In a 2.00 mL reaction mix, the final concentrations are 25 mM sodium acetate, 0.5% (w/v) starch and 0.15–0.60 unit of amyloglucosidase.
Enzymatic Stop Reaction
1. Pipette the following reagent into suitable containers:
ReagentTest 1 (mL)Test 2 (mL)Test 3 (mL)Blank (mL)
Starch Solution1.001.001.001.00
2. Equilibrate to 55 °C. Then add:
ReagentTest 1 (mL)Test 2 (mL)Test 3 (mL)Blank (mL)
Enzyme Solution0.500.701.00
3. Immediately mix by swirling and incubate at 55 °C for exactly 3 minutes. Then add:
ReagentTest 1 (mL)Test 2 (mL)Test 3 (mL)Blank (mL)
TCA Solution0.300.300.300.30
Enzyme Solution0.500.301.00
4. Mix by swirling and adjust to pH 7.0 with solid Sodium bicarbonate (S8875).
5. Filter through a 0.1 µM PVDF syringe filter (F7523) and use Filtrate in Enzymatic Activity Determination, step 4.
Enzymatic Activity Determination
1. Pipette the following reagent into suitable cuvettes:
ReagentTest 1 (mL)Test 2 (mL)Test 3 (mL)Blank (mL)
Glucose Assay Reagent2.802.802.802.80
2. Equilibrate to 25 °C. Monitor the A340 until constant, using a suitably thermostatted spectrophotometer.
3. Record the initial A340 for both the Tests and the Blank.
4. Then add:
ReagentTest 1 (mL)Test 2 (mL)Test 3 (mL)Blank (mL)
Appropriate Test Filtrate, Enzymatic Stop Reaction, step 50.200.200.20
Blank Filtrate, Enzymatic Stop Reaction, step 50.20
5. Immediately mix by inversion. Monitor the A340 until the DA340/min is <0.0020 and this rate is maintained for at least 5 minutes.
6. Record the final A340 for both the Tests and the Blank.
Results
Calculations
1. ΔA340 = A340 Final – A340 Initial
2.
Units/mL enzyme =(ΔA340 Test – ΔA340 Blank) (180) (2.3) (3.0) (df)
(6.22) (1,000) (mL Enzyme) (0.20)
where:
180 = Micrograms of glucose per micromole of glucose
2.3 = Total volume (in milliliters) of Enzymatic Stop Reaction, steps 1–5
3.0 = Total volume (in milliliters) of Enzymatic Activity Determination, steps 1–6
df = Dilution factor
6.22 = Millimolar extinction coefficient of β-NADPH at 340 nm
1,000 = Conversion factor from micrograms to milligrams         
ml Enzyme = Volume of Enzyme Solution added in Enzymatic Stop Reaction, step 2
0.20 = Volume (in milliliters) from Enzymatic Stop Reaction used in Enzymatic Activity Determination
3.
Units/mg solid =units/mL enzyme
mg solid/mL enzyme
Materials
Loading
03/14-1
References
1.
1963. Bergmayer, H. U.: Methods of Enzymatic Analysis. Verlag Chemie G.m.b.H., Weinheim/Bergstr. (Germany) and Academic Press, New York and London. 1963, XXIV, 1064 pages with 78 figures, 4 colored diagrams and 41 tables. Clothbound DM 120. ? = $ 30.00. Starch/Stärke. 15(7):272-272. https://doi.org/10.1002/star.19630150713