Enzymatic Assay of Elastase (EC 3.4.21.36)
where:
SucAla3-pNA = N-Succinyl-Ala-Ala-Ala-p-nitroanilide
SucAla3 = N-Succinyl-Ala-Ala-Ala
pNA = p-Nitroanilide
Unit Definition – One unit of Elastase will hydrolyze 1.0 µmole of N-succinyl-L-Ala-Ala-Ala-p-nitroanilide per minute at pH 8.0 at 25 °C.
Precautions
Please consult the Safety Data Sheet for information regarding hazards and safe handling practices.
Reagents and Equipment Required
Trizma® base (Product No. T1503)
N-Succinyl-Ala-Ala-Ala-p-nitroanilide (Product No. S4760)
Preparation Instructions
Use ultrapure water (≥18 MΩxcm resistivity at 25 °C) for the preparation of reagents.
Buffer (100 mM Tris HCl, pH 8.0 at 25 °C) – Prepare a 12.1 mg/mL solution of Trizma® base (Product No. T1503) in ultrapure water. Adjust the pH to 8.0 at 25 °C with 1 M HCl.
Substrate Solution (4.4 mM SucAla3-pNA Solution) – Prepare 2 mg/mL solution of N-Succinyl-Ala-Ala-Ala-p-nitroanilide (Product No. S4760) in Buffer.
Enzyme Solution (Elastase) – Immediately before use, prepare a solution containing 0.2–0.5 unit/mL of Elastase in cold (2–8 °C) buffer.
Procedure
In a 3.00 mL reaction mix, the final concentrations are 96.7 mM Trizma®, 0.29 mM N-Succinyl-Ala-Ala-Ala-p-nitroanilide, and 0.02–0.05 unit of Elastase.
1. Pipette the following reagents into suitable cuvettes:
Reagent | Test (mL) | Blank (mL) |
---|---|---|
Buffer | 2.70 | 2.80 |
Substrate Solution | 0.20 | 0.20 |
2. Mix by inversion and equilibrate to 25 °C. Then add:
Reagent | Test (mL) | Blank (mL) |
---|---|---|
Enzyme Solution | 0.10 | – |
1. | |
---|---|
Units/mL enzyme = | (ΔA410/min Test – ΔA410/min Blank) (3.00) (df) |
(8.8) (0.10) |
where:
3.00 = Total volume (mL) of assay
df = Dilution factor
8.8 = Millimolar extinction coefficient of p-Nitroaniline at 410 nM at pH 8.0
0.1 = Volume (mL) of Enzyme Solution used
2. | |
---|---|
Units/mg solid = | units/mL enzyme |
mg solid/mL enzyme |