Merck
HomeChemical Analysis for Food and Beverage TestingAnalysis of Antioxidants in Bakery Products
Analysis of Antioxidants in Bakery Products following the GB 5009.32-2016 Method
Dean Duan, Senior Scientist
Merck R&D APAC Lab, Shanghai, China
Abstract
The Discovery® HS C18 column was used to separate the 9 antioxidants propyl gallate (PG), 1-(2,4,5-trihydroxyphenyl)-1-bunanone (THBP), tert-butylhydroquinone (TBHQ), nordihydroguaiaretic acid (NDGA), butylated hydroxyanisole (BHA), 3,5-di-tert-butyl-4-hydroxybenzyl alcohol (Ionox-100), octyl gallate (OG), butylated hydroxytoluene (BHT) and dodecyl/lauryl gallate (DG) in pastries following the Chinese national standard GB 5009.32-2016 method for determination of 9 antioxidants in food. The here-developed method using a Discovery® HS C18 column fulfills the system suitability criteria of the GB 5009.32-2016 method and exceeds the sensitivity criteria.
Section Overview
Introduction
Food antioxidants are additives that prevent or delay the oxidative deterioration of food to improve its stability and extend its storage period. Within the permitted levels for normal consumption, these antioxidants do not pose a health risk. However, individuals who are allergic or have asthma may develop hypersensitive reactions over time.1 The permissible limits for antioxidants vary for different foods in the different countries.2
The Chinese national standard GB 5009.32-2016 method3 describes a C18 HPLC column for the antioxidant measurement. Following this, a Discovery® HS C18 column was used for the here described analysis of 9 antioxidants (Figure 1), propyl gallate (PG), 1-(2,4,5-trihydroxyphenyl)-1-bunanone (THBP), tert-butylhydroquinone (TBHQ), nordihydroguaiaretic acid (NDGA), butylated hydroxyanisole (BHA), 3,5-di-tert-butyl-4-hydroxybenzyl alcohol (Ionox-100), octyl gallate (OG), butylated hydroxytoluene (BHT) and dodecyl/lauryl gallate (DG), in a bread roll sample.
Chemical structures of aminoglycoside antibiotics in two rows. The first row shows the structures of propyl gallate (PG), 1-(2,4,5-trihydroxyphenyl)-1-bunanone (THBP), tert-butylhydroquinone (TBHQ), nordihydroguaiaretic acid (NDGA), and butylated hydroxyanisole (BHA) from left right. The second row shows Ionox-100, octyl gallate (OG), butylated hydroxytoluene (BHT) and dodecyl/lauryl gallate (DG) from left to right.
Figure 1.Structures of food antioxidant compounds investigated in this study.
Experimental
Standard Preparation
  • Antioxidants stock solution (1 mg/mL each): Weigh 10 mg of each of the 9 antioxidants into a 10 mL brown volumetric flask. Add ~8 mL of acetonitrile and sonicate for 5 minutes. Top-up to mark with acetonitrile and mix well.
  • Standard solutions for external calibration: Dilute stock solution to 1, 2, 5, 10, 20, 50, 100 and 200 µg/mL with acetonitrile.
Sample Preparation
Saturated solvent solutions: Take 200 mL of acetonitrile and 200 mL hexane into a 500 mL separating funnel. Cap and shake the funnel vigorously for 1 minute and allow it to stand for 5 minutes. Separate the solvents by draining the lower layer (hexane saturated acetonitrile) into a beaker followed by the upper layer (acetonitrile saturated hexane) into another beaker.
  • Cut the pastry sample into smaller pieces and grind it into a paste or powder.
  • Weigh 1 g (accurate to 0.01 g) of sample into a 50 mL centrifuge tube. For spiked samples, add appropriate quantities of the mixed antioxidants stock solution to the sample and vortex for 1 minute.
  • Add 5 mL of a hexane solution saturated with acetonitrile to the sample, vortex for 1 minute, and stand for 10 minutes.
  • Add 5 mL of saturated sodium chloride solution to the above.
  • Add 5 mL of acetonitrile solution saturated with hexane to the above. Vortex for 2 minutes and centrifuge at 3000 rpm for 5 minutes. Transfer the acetonitrile layer (middle layer) to a 150 mL distillation flask.
  • Repeat extraction twice with a 5 mL acetonitrile solution saturated with hexane. Collect all extracts into the same distillation flask.
  • Evaporate the extract to dryness using a rotary evaporator under nitrogen in a 40 °C water bath.
  • Dissolve the residue in 2 mL of acetonitrile. Filter through a 0.22 μm membrane before HPLC analysis (Table 1).
HPLC-UV Analysis
LC Conditions
Column:
Discovery® HS C18, 5 μm, 25 cm × 4.6 mm I.D. (568523-U)
Mobile phase:
[A] 0.5% Formic acid in water; [B] methanol
Gradient:
Time (min)
A%
B%
0.00
50.0
50.0
5.00
50.0
50.0
15.00
20.0
80.0
20.00
20.0
80.0
25.00
10.0
90.0
27.00
50.0
50.0
30.00
50.0
50.0
Flow rate:
1.0 mL/min
Pressure:
2550 psi
Column temp.:
35 °C
Detector:
UV, 280 nm
Injection:
10 µL
Sample:
Acetonitrile extract and standards
Table 1. HPLC conditions used for antioxidant analysis
Method Suitability, System Suitability, Calibration
Acceptance Criteria as described in GB 5009.32-2016
  • Linear correlation coefficient: R2 > 0.99
  • Method recovery: 80 to 120%
  • LOQ <20 mg/kg
Results & Discussion
Chromatographic results for a standard injection, a pastry sample and a spike pastry sample are displayed in the Figures 2-4. The specificity/retention times of the 9 antioxidants is shown in Table 2. The calibration curve for propyl gallate (PG) is displayed as example in the Figure 5. Chromatographic data of the pastry sample spiked with 9 antioxidants at 20 mg/kg (Figure 4) is shown in Table 3. The repeatability (n=5) for a standard solution at 20 µg/mL each compound was between 0.19 and 0.51 %RSD (Table 4). The external calibration linearity data from 1 to 200 µg/mL showed R2 values >0.999 (Table 5). The % recovery stayed withing the required 80-120% range (Table 6). Table 7 compares the LOD and LOQ values of this method and the GB 5009.32-2016 method, showing the developed method exceeds the criteria. The determination of antioxidants with an actual bread roll sample sourced from a local store is shown in Table 8. In the investigated unspiked sample a BHA content of 4.82 mg/kg was determined.
HPLC-UV chromatogram of a mixed standard solution consisting of 9 antioxidants. Intensity (mAU) on the y-axis and retention time (minutes) on the x-axis. Major ticks on x-axis at 5, 10, 15, 20, 25, and 30 minutes, and on y-axis at 10, 20, 30, 40, 50, 60, 70, 80, 90, and 100. The green plot initially runs parallel to the X-axis, shows peaks of varying heights labeled 1 to 9 from left to right, starting with PG at 5.9 minutes, THBP at 9 minutes, TBHQ at 9.6 minutes, NDGA at 15.2 minutes, BHA at 16.2 minutes, Ionox-100 at 18.1 minutes, OG at 19 minutes, BHT at 28 minutes, and DG at 28.4 minutes.
Figure 2.Mixed standard solution of 9 antioxidants, 20 μg/mL. For Peak IDs see Table 2.
Peak ID
Compound
Abbreviation
Retention Time (min)
RRT
Resolution
Tailing Factor
1
Propyl gallate
PG
5.9
1.00
--
0.86
2
1-(2,4,5-Trihydroxyphenyl)-1-bunanone
THBP
9.0
1.53
10.7
0.97
3
tert-Butylhydroquinone
TBHQ
9.6
1.63
2.1
0.97
4
Nordihydroguaiaretic acid
NDGA
15.2
2.59
27.9
0.96
5
Butylated hydroxyanisole
BHA
16.2
2.75
6.7
0.94
6
3,5-Di-tert-butyl-4-hydroxybenzyl alcohol
Ionox-100
18.1
3.08
14.2
0.94
7
Octyl gallate
OG
19.0
3.23
6.5
0.96
8
Butylated hydroxytoluene
BHT
28.0
4.76
51.7
0.93
9
Dodecyl gallate
DG
28.4
4.82
1.8
0.94
Table 2.Chromatographic data of the 9 antioxidants standard solution at 20 μg/mL
HPLC-UV chromatogram of an unspiked pastry sample. Intensity on the y-axis and retention time (minutes) on the x-axis. Major ticks on x-axis at 10, 20, and 30 minutes, and on y-axis at 10, 20, 30, 40, 50, 60, 70, 80, 90, and 100. The plot initially run parallel to the x-axis at then shows small height overlapping peaks between 2 to 5 minutes. This is followed by a large peak at 7 minute and then a very small peak labeled 5 around 16 minute, being BHA.
Figure 3.Chromatogram of an unspiked pastry sample (bread roll).
HPLC-UV chromatogram of a spiked bread roll sample with 9 antioxidants at 20 mg/kg. Intensity on the y-axis and retention time (minutes) on the x-axis. Major ticks on x-axis at 5, 10, 15, 20, 25 and 30 minutes, and on y-axis at 10, 20, 30, 40, 50, 60, 70, 80, 90, and 100. The plot initially run parallel to the x-axis at 20 mAU and then shows a few unlabeled peaks and 9 distinct labeled peaks from 1-9 along left to right direction.
Figure 4.Spiked pastry sample (bread roll) with 9 antioxidants at 20 mg/kg.
Peaks
Compound
Retention Time (min)
Resolution
Tailing Factor
1
PG
5.9
5.5
0.87
2
THBP
9.0
6.0
0.95
3
TBHQ
9.5
2.1
0.95
4
NDGA
15.2
27.6
0.95
5
BHA
16.2
5.8
0.95
6
Ionox-100
18.1
12.4
0.92
7
OG
19.0
6.3
0.94
8
BHT
27.9
17.0
0.92
9
DG
28.3
1.7
0.93
Table 3.Chromatographic data of pastry sample spiked with the 9 antioxidants at 20 mg/kg
Calibration Data
Calibration in the range of 1-200 µg/mL with 8 standard solutions provided linear calibration curves with R2 values between 0.99958 and 0.99996 (Table 5) meeting the method requirement of R2 > 0.99. As example, the curve for PG is given in Figure 5.
Compound
Range (µg/mL)
Linear equation
R2
PG
1 - 200
y = 24,922.61009x - 11,621.71451
0.99991
THBP
1 - 200
y = 37,473.03828x - 50,168.98139
0.99969
TBHQ
1 - 200
y = 8,268.98088x - 6,319.57287
0.99994
NDGA
1 - 200
y = 11,848.51640x - 11,774.29529
0.99958
BHA
1 - 200
y = 7,790.82192x - 4,247.73808
0.99992
Ionox-100
1 - 200
y = 2,896.25484x - 2,062.23462
0.99980
OG
1 - 200
y = 20,953.38489x - 15,011.41737
0.99979
BHT
2 - 200
y = 4,156.59672x - 3,457.70460
0.99988
DG
1 - 200
y = 16,748.24953x - 4,498.60232
0.99996
Table 5.External calibration linearity of 9 antioxidants, 8 calibrants each (except BHT with 7 calibrants)
Intensity v/s concentration measured in µg/mL plot representing calibration curve obtained for propyl gallate at 8 concentrations between 1 - 200 µg/mL. Major ticks on x-axis at 40, 80, 120, 160, and 200 µg/mL, and on y-axis at 20, 40 and 60 * sec × 100000 mAU. The linear equation obtained is y = 24,922.61009x - 11,621.71451 with R² = 0.99991.
Figure 5.Calibration curve of PG shown as example.
Repeatability
The repeatability for the 9 analytes was between 0.19 to 0.51 %RSD (n=5) for the 20 µg/mL standard solution (Table 5).
Compound
Mean Area
Std Dev
% RSD
PG
486621
1405
0.29
THBP
726323
3197
0.44
TBHQ
156435
796
0.51
NDGA
228485
745
0.33
BHA
152404
307
0.20
Ionox-100
56983
108
0.19
OG
411556
1551
0.38
BHT
80597
308
0.38
DG
325426
1323
0.41
Table 4.Repeatability (n=5) of 9 antioxidants standard solution at 20 µg/mL
Recovery Data    
For spiked samples (20 µg/kg) for the 9 analytes, recovery ranged from 77.2 to 115.4% and %RSD for triplicate measurement from 1.40 to 4.12 % (Table 6). Regarding the method criteria for recovery (80 to 120%) only TBHQ showed with 77.2% a recovery slightly below the limit.
Compound
Recovery (%)
% RSD (n=3)
PG
101.1
2.30
THBP
107.9
1.40
TBHQ
77.2
2.80
NDGA
98.0
0.55
BHA
115.4
2.00
Ionox-100
106.6
1.90
OG
90.8
3.30
BHT
85.7
4.12
DG
80.8
2.02
Table 6.% recovery for 9 antioxidants upon cleanup extraction for a bread roll sample spiked at 20 mg/kg
Method Sensitivity
The method showed good sensitivities exceeding the requirement of the GB method (Table 7). Determined LODs were <1.1 mg/kg and LOQs of <3.07 mg/kg, exceeding the methods criteria of 20 mg/kg.
Developed Method
GB requirement
Compound
LOD (mg/kg)
LOQ (mg/kg)
LOD (mg/kg)
LOQ (mg/kg)
PG
0.93
2.80
<2
<20
THBP
1.01
3.07
<4
<20
TBHQ
0.55
1.68
<10
<20
NDGA
0.46
1.39
<4
<20
BHA
0.35
1.07
<10
<20
Ionox-100
0.68
2.06
<20
<20
OG
0.33
1.00
<2
<20
BHT
0.80
2.42
<4
<20
DG
0.48
1.44
<10
<20
Table 7.LOD and LOQ of method against GB method requirements
Sample measurement
For the investigated unspiked pastry sample only BHA was detected at 4.82 mg/kg (Table 8).
Compound
Amount (mg/kg)
PG
<LOD
THBP
<LOD
TBHQ
<LOD
NDGA
<LOD
BHA
4.82
Ionox-100
<LOD
OG
<LOD
BHT
<LOD
DG
<LOD
Table 8.Results of antioxidants detected in the unspiked pastry sample
Conclusion
The Discovery® HS C18 column could separate the 9 antioxidants in the pastry matrix efficiently with excellent peak shapes. The shown method meets the acceptance criteria of GB 5009.32-2016 regarding recovery (except for TBHQ) and exceeds the sensitivity requirement with lower LOD and LOQ values.
See more applications for Food & Beverage testing.
Related Products
Loading
References
1.
Food Safety Focus: Should We Worry about Preservatives and Antioxidants in Food – Or Should We Not?. [Internet]. Centre for Food Safety, The Government of the Hong Kong Special Administrative Region.[cited 31 Dec 2022]. Available from: https://www.cfs.gov.hk/english/multimedia/multimedia_pub/multimedia_pub_fsf_199_02
2.
WHO Food Additives Series: 42. [Internet]. WHO INCHEM. International Programme on Chemical Safety– Safety Evaluation of Certain Food Additives. .[cited 31 Dec 1998]. Available from: https://www.inchem.org/documents/jecfa/jecmono/v042je24.htm
3.
Determination of nine antioxidants. [Internet]. GB 5009.32-2016 .[cited 31 Dec 2022]. Available from: https://www.gbstandards.org/China_standard_english.asp?code=GB%205009.232-2016