MilliporeSigma
HomeChemical Analysis for Food and Beverage TestingAnalysis of Caffeine and Catechins in Green Tea
HPLC-UV Analysis of Caffeine and Catechins in Green Tea (Decaffeinated)
Dr. Ajay Kaparwan
Mumbai Application Laboratory, India
INTRODUCTION
Tea from the leaves of Camellia sinensis is used to make green tea, black tea, white tea, and oolong tea. The different teas are possible due to the different processing employed. Green tea is not fermented so its polyphenolic components of which the majority are catechins are not oxidized. As such, this decaffeinated green tea assay focuses on catechins and caffeine.
Here, we describe and validate an HPLC method as per Ph. Eur. Monograph 2668 (04/2018)1 as a reference method for the separation and quantification of catechins and caffeine in decaffeinated green tea using a Chromolith® High Resolution RP-18e 150 x 4.6 mm column (Table 1). The monolithic silica column, due to its bimodal pore structure, provides advantages regarding column backpressure and matrix robustness for the analysis of such natural matrix samples.
Chemical structures of catechins and caffeine in two rows. The first row from left to right shows catechin, epicatechin, epigallocatechin, and epigallocatechin-3-O-gallate. The second row from left to right shows, gallocatechin, epicatechin-3-O-gallate, gallocatechin-3-O-gallate, and caffeine.
Figure 1.Chemical structures of catechins and caffeine
EXPERIMENTAL CONDITIONS
Column:
Chromolith® High Resolution RP-18e, 150 x 4.6 mm (1.52023)
Mobile Phase:
[A] 0.01% v/v formic acid prepared by adding 0.1 mL to 1000 mL water;
[B] 0.01% v/v formic acid prepared by adding 0.1 mL to 1000 mL methanol;
[C] acetonitrile;
All mobile phases were filtered through a 0.45 µm membrane filter.
Gradient:
Time
A(%)
B(%)
C(%)
  0
97
  0
3
  5
97
  0
3
23
67
30
3
29
67
30
3
30
30
67
3
31
30
67
3
33
97
  0
3
37
97
  0
3
Flow rate:
0.5 mL/min
Back pressure:
40 bar (576 psi)
Column temp.:
41° C
Detector:
UV @ 210 nm
Injection:
2 µL
Sample(s) & Sample Preparation
Extraction solution:
Water:Methanol (30:70 v/v) at 70°C.
Stabilizing solution:
Weigh 250 mg of (ethylenedinitrilo) tetra-acetic acid and 250 mg of ascorbic acid into a 1000 mL volumetric flask. Add 500 mL water and mix to dissolve. Add 100 mL of acetonitrile and top to mark with water.
Test solution:
Weigh 0.2 g of finely crushed decaffeinated green tea sample into a test tube. Add 5 mL of extraction solution to this and stopper it. Vortex to mix and place this in a 70°C water bath. At 5 min, remove from bath and vortex. Return to bath and after 10 min, remove to vortex again and allow to cool. Centrifuge at 2500 rpm for 10 minutes. Decant the supernatant to 10 mL of a volumetric flask. Repeat the extraction with a 5 mL extraction solution and combine the extracts into the 10 mL flask. Top up with cold extraction solution and mix well. Further, dilute 1 mL of this solution to 20 mL with stabilizing solution before filtering through a 0.45 µm PVDF syringe filter.
Catechin standards:
Weigh 7 mg of the catechin into a 20 mL volumetric flask. Add diluent and swirl, sonicate for 20 minutes. Dilute 920 µL of this solution to 10 mL with diluent. Final conc. is 32 µg/mL.
Diluent: 5 mL of extracting solution to 100 mL volumetric flask and top to mark with stabilizing solution. Mix well and sonicate for 5 minutes before use.
Reference solution A:
Weigh 20 mg of Epigallocatechin-3-O-gallate RS and 20 mg of Caffeine RS into a 25 mL volumetric flask. Add water to dissolve and top to mark. Dilute 2 mL of this solution to 50 mL with stabilizing solution. Filter through 0.45 µm PVDF syringe filter. Final conc. is 32 mg/mL.
RESULTS
Chromatogram of blank sample plotted between intensity and retention time with a peak at 4.9 minutes
Figure 2.Blank chromatogram.
Chromatogram of reference solution A plotted between intensity and retention time showing two peaks labeled Epigallocatechin-3-O-gallate and Caffeine
Figure 3.Chromatogram of reference solution A.
Chromatogram of catechin standard mixture plotted between intensity and retention time showing different labeled peaks of catechins and caffeine
Figure 4.Chromatogram of catechin and caffeine standard mixture. See Table 4 for retention times (determined with single solution standards).
Chromatogram of green tea sample 1 plotted between intensity and retention time showing labeled peaks of gallocatechin, epigallocatechin, catechin, caffeine, epigallocatechin-3-O-gallate, epicatechin, gallocatechin-3-O-gallate, and epicatechin-3-O-gallate
Figure 5.Chromatogram of green tea sample 1
Chromatogram of green tea sample 2 between intensity and retention time showing labeled peaks of gallocatechin, epigallocatechin, catechin, caffeine, epigallocatechin-3-O-gallate, epicatechin, gallocatechin-3-O-gallate, and epicatechin-3-O-gallate
Figure 6.Chromatogram of green tea sample 2.
Sr. No.
Green Tea
% Caffeine*     as Per EP Monograph
% Catechin expressed as Epigallocatechin-3-O-gallate as per EP Monograph
% Water Content by KF
1
Sample 1
0.17
10.66
8.2
2
Sample 2
0.09
6.25
10.5
Table 2.Results for 2 green tea samples
*Decaffeinated tea would still have minor amounts of caffeine as observed in the tea sample chromatograms. The decaffeination process can reduce the caffeine amount to <2.5% of the original caffeine load.
System Suitability and Repeatability Data
Area in (mAU)
Standard
Gallocatechin
Epigallocatechin
Catechin
Caffeine
Epigallocatechin-3-O-Gallate
Epicatechin
Gallocatechin-3-O-Gallate
Epicatechin-3-O-Gallate
Standard_1
1437880
1458772
1221835
842610
1528633
1238253
1499736
991214
Standard_2
1433198
1471379
1232416
849415
1541607
1247795
1511801
999639
Standard_3
1447688
1489368
1248150
861672
1562190
1263432
1537666
1012880
Standard_4
1438758
1459020
1222586
845011
1530189
1238082
1501816
992558
Standard_5
1444325
1472992
1233040
850780
1543571
1249197
1518150
1000504
Mean
1440369.8
1470306.2
1231605
849897.6
1541238
1247351.8
1513833.8
999359
Standard Deviation
5688.40
12571.12
10644.83
7359.28
13467.85
10378.96
15279.98
8613.96
% RSD
0.4
0.9
0.9
0.9
0.9
0.8
1
0.9
Table 3.Reproducibility of area counts (mAU) of catechin and caffeine standard (32 µg/mL each compound)
Retention Time (minutes)
Standard
Gallocatechin
Epigallocatechin
Catechin
Caffeine
Epigallocatechin-3-O-Gallate
Epicatechin
Gallocatechin-3-O-Gallate
Epicatechin-3-O-Gallate
Standard_1
8.516
15.037
15.955
17.949
19.911
20.466
21.844
24.605
Standard_2
8.511
15.033
15.954
17.95
19.914
20.468
21.847
24.61
Standard_3
8.512
15.036
15.956
17.954
19.919
20.474
21.853
24.615
Standard_4
8.507
15.029
15.948
17.946
19.906
20.461
21.84
24.603
Standard_5
8.512
15.046
15.964
17.963
19.923
20.479
21.858
24.621
Mean
8.5116
15.0362
15.9554
17.9524
19.9146
20.4696
21.8484
24.6108
Standard Deviation
0.00321
0.00630
0.00573
0.00658
0.00666
0.00702
0.00716
0.00736
% RSD
0.04
0.04
0.04
0.04
0.03
0.03
0.03
0.03
Table 4.Retention Time of Caffeine and Catechin standards (32 ug/mL)
Component
Ph. Eur.  Specification
Observed Value
% RSD
2%
<2%
Retention Time
1%
<1%
Tailing Factor
2
<2
Resolution, Reference Solution A
Minimum 1.5
>2
Table 5.System Suitability Criteria set in Ph. Eur and performance observed
Method Sensitivity and Linearity
Compound
STYX
Slope
LOD (µg/mL)
LOQ (µg/mL)
Correlation Coefficient
Gallocatechin
57107.43385
41551.7194
4.54
13.74
0.997
Epigallocatechin
54498.80895
43955.46957
4.09
12.4
0.998
Catechin
52338.64729
38664.96098
4.47
13.54
0.997
Caffeine
33376.35649
24990.34237
4.41
13.36
0.997
Epigallocatechin-3-O-gallate
36394.676
46555.82273
2.58
7.82
0.999
Epicatechin
38574.65905
38499.92522
3.31
10.02
0.998
Gallocatechin-3-O-gallate
47867.18528
47943.76143
3.29
9.98
0.998
Epicatechin-3-O-gallate
31694.71884
31686.72165
3.3
10
0.998
Table 6.Determined LOD, LOQ and linearity of method using 10 standard solutions spanning the concentration range 0.08 – 48 µg/mL for each compound.
Calibration curves plotted between intensity and concentration (µg/mL) obtained for caffeine, epigallocatechin-3-O-gallate (purple color), gallocatechin-3-O-gallate (pink color), epigallocatechin (blue color), gallocatechin (dark purple color), catechin (light green color), epicatechin (dark green color), epicatechin-3-O-gallate (dark blue color), and caffeine (yellow color) with equations of the form y=mx+c for each of the obtained lines.
Figure 7.Calibration curves for the catechins and caffeine.
% Recovery values of the Catechins and Caffeine spiked into decaffeinated green tea sample are displayed in Table 7.
No.
Spike Level
Added (µg/mL)
Found (µg/mL)
Recovery (%)
Gallocatechin
1
LOQ
14
13.6
97.6
2
50%
16
16.9
105.9
3
100%
32
31.2
97.6
4
150%
48
51.6
107.5
Epigallocatechin
1
LOQ
13
11.8
91.1
2
50%
16
16.1
100.6
3
100%
32
29.4
92
4
150%
48
47.6
99.1
Catechin
1
LOQ
14
13.5
96.5
2
50%
16
14.6
91.2
3
100%
32
34
106.4
4
150%
48
47.6
99.1
Caffeine
1
LOQ
14
13.8
98.3
2
50%
16
14.4
90.1
3
100%
32
30.2
94.3
4
150%
48
47.9
99.7
Epigallocatechin-3-O-Gallate
1
LOQ
8
7.7
96.4
2
50%
16
15.6
97.3
3
100%
32
30.6
95.5
4
150%
48
45.8
95.4
Epicatechin
1
LOQ
10
9.2
91.9
2
50%
16
14.5
90.6
3
100%
32
31.3
97.8
4
150%
48
51.8
107.9
Gallocatechin-3-O-Gallate
1
LOQ
10
10.2
101.5
2
50%
16
15.1
94.5
l3
100%
32
33.5
104.8
4
150%
48
45.9
95.7
Epicatechin-3-O-Gallate
1
LOQ
10
9.1
91.3
2
50%
16
16.3
102.1
3
100%
32
30.2
94.2
4
150%
48
50.5
105.1
Table 7.Recovery levels at different spike levels
CONCLUSION
A gradient reverse phase HPLC method for the green tea assay using the monolithic HPLC column following the Ph. Eur. Monograph 2668 (04/2018)has been modified and validated. It was shown that the Chromolith® High resolution column offers good resolution (>2) between each of the catechins present in decaffeinated green tea.
The assay method as per Ph. Eur. (edition 10) is simple, linear, and specific. The Limits of Detection (LOD) ranged from 2.58 to 4.54 µg/mL and the Limits of Quantification (LOQ) was from 7.82 to 13.74 µg/mL with good linearity up to 48 µg/mL for the determined catechins and caffeine.
The % RSD for system precision of the catechins and caffeine was <2%. The analyte recovery of the catechins and caffeine ranged from 90.1 to 107.9%.
The Chromolith® High Resolution RP-18e column is an ideal column for this application as it offers due to its bimodal pore structure the advantage of low back pressure and matrix robustness for a longer lifetime compared to the traditional porous particulate columns without sacrificing resolution, accuracy, and precision.
RELATED PRODUCTS
Loading
REFERENCES
1.
Ph. Eur. (edition 10) 2668 (04/2018) GREEN TEA Camelliae sinensis non fermentata folia. [Internet].