MilliporeSigma
HomeCustom siRNA Duplex Plates
Custom siRNA Duplex Plates
We offer custom siRNA duplexes supplied in 96-well plates for high throughput screening applications. The siRNAs can be chemically modified to your specifications. All siRNAs are analyzed by mass spec prior to release.
Custom siRNA Duplex Plates Specifications
Quantities0.2 OD (1 nmol), 0.4 OD (2nmol), 2 OD (10 nmol)
PurificationDesalt, HPLC (2 OD)
Sequence Length19 to 27mer duplexes
Modifications1 and 2 nmol = None
10 nmol = Biotin, Phosphate, Amino-Modifier (C6, C7), 6-FAM™, Cyanine 3, Cyanine 5, Cyanine 5.5, 2’- OMe, Phosphorothioate, Flipped Base, 2’-Fluoro, Inverted Abasic, Thiol-Modifier-C6 S-S, etc.
Inquire for others at sirnarequest@milliporesigma.com
Quality Control100% mass spectrometry**
FormatSupplied dry in plates
Minimum Order size24 duplexes
**Depending on manufacturing site, PAGE may be used to assess siRNA duplexes
If you have needs that are different from the other general specifications presented, please send a request to [email protected].
MISSION is a trademark of Merck KGaA, Darmstadt, Germany and/or its affiliates. Label License.