MilliporeSigma
Glycogen Synthase Kinase 3 (GSK-3) Overview
Glycogen synthase kinase 3 (GSK-3) is a highly conserved family of protein kinases. In humans, two genes encode two distinct, but closely related GSK-3 forms, referred to as GSK-3α and GSK-3β. They display 84% overall identity and 98% identity within their catalytic domain, the main difference between the two coming from an extra Gly-rich stretch in the N-terminal domain of GSK-3α. Yet they are not functionally interchangeable as demonstrated by the embryonic lethal phenotype of GSK-3β knock-outs. GSK-3β2 is an alternate splicing variant of GSK-3ßβ expressed in brain.
GSK-3β has been crystallized with various ligands. Its overall shape is shared by all kinases, and comprises a small, N-terminal lobe consisting mostly of β sheets and a large, C-terminal lobe, essentially formed of α-helices. The ATP-binding pocket is located between the two lobes. Arg96, Arg180 and Lys205 form a small pocket where the phosphate group of the primed substrate and the pseudo-substrate binds.
GSK-3α/β is regulated by phosphorylation on Ser21/9 (inhibitory) and on Tyr279/216 (activating). The phospho-Ser21/9 N-terminal domain of GSK-3 acts as a pseudo-substrate blocking access of substrates to the catalytic site. The T-loop domain containing unphosphorylated Tyr279/216 is thought to prevent access of substrates to the catalytic site, and Tyr phosphorylation to release this inhibition. GSK-3 is also regulated by interactions with other proteins. Axin and presenilin act as docking proteins that allow the substrate to interact with the priming kinase. The substrate indeed requires a priming phosphorylation by another kinase on a residue located C-terminal to the GSK-3 phosphorylation site. The substrate recognition site of GSK-3 is -S-X-X-X-Sp-, where Sp is the priming pre-phosphorylated Ser. GSK-3β is a predominantly cytoplasmic kinase, but it is also present in the nucleus and in mitochondria. FRAT-1 promotes the nuclear efflux of GSK-3β. LANA, a Kaposi virus protein, sequesters GSK-3β in the nucleus.
GSK-3 plays a major function in Wnt signalling pathways. When the Wnt pathway is stimulated, GSK-3 is inactivated, β-catenin builds up and accumulates in the nucleus where it forms with TCF/LEF, a transcription factor regulating a large variety of genes. GSK-3 phosphorylates the cell cycle regulators β-catenin, cyclin D1, cyclin E, p21CIP1 and c-Myc, leading to their ubiquitin-dependent destruction. In the absence of insulin, active GSK-3 phosphorylates and inactivates glycogen synthase and eIF2B. Binding of insulin to its plasma membrane receptor leads to PKB/AKT activation, resulting in phosphorylation and inactivation of GSK-3. Consequently, glycogen synthase and eIF2B are activated, and glycogen and protein synthesis are stimulated. GSK-3 plays a pro-apoptotic function in neuronal cells. In contrast, GSK-3 is necessary also for cell survival as demonstrated by the massive TNFα-induced hepatocyte apoptosis which leads to death in early embryonic GSK-3β knockout mice. GSK-3 activity/inactivity or spatial distribution play an essential role during development (polarity determination). Finally, GSK-3 and CK1 regulate the circadian clock in Drosophila and in mammals.
Five sets of data have stimulated altogether the search for pharmacological inhibitors of GSK-3: (i) the mood stabilizing properties of lithium, the first GSK-3 inhibitor to be described; (ii) the insulin-mimetic properties of GSK-3 inhibitors; (iii) the interaction of GSK-3 with presenilin-1, the GSK-3-dependent amyloid-β production and abnormal tau phosphorylation in Alzheimer's disease; (iv) the involvement of GSK-3 in neuronal cell death and the neuroprotection provided by GSK-3 inhibitors following various insults; and (v) the maintenance of pluripotency of embryonic stem cells in the absence of feeder cells by GSK-3 inhibitors. Over 30 GSK-3 inhibitors have been identified, among which seven have been co-crystallized with GSK-3β, all of which localize within the ATP-binding pocket of the enzyme. GSK-3 inhibitors are thus evaluated on Alzheimer's disease and other neurodegenerative diseases, bipolar affective disorders, diabetes and diseases caused by unicellular parasites.
The Table below contains accepted modulators and additional information. For a list of additional products, "Similar Products" section below.
Abbreviations
SB 216763: 3-(2,4-Dichlorophenyl)-4-(1-methyl-1H-indol-3-yl)-1H-pyrrole-2,5-dione
SB 415286: 3-[(3-Chloro-4-hydroxyphenyl)amino]-4-(2-nitrophenyl)-1H-pyrrol-2,5-dione
Similar Products
Loading
References
1.
S. A, R.H. B, G. A. 2011. The Possible Involvement of Glycogen Synthase Kinase-3 (GSK-3) in Diabetes, Cancer and Central Nervous System Diseases. CPD. 17(22):2264-2277. https://doi.org/10.2174/138161211797052484
2.
Bertrand J, Thieffine S, Vulpetti A, Cristiani C, Valsasina B, Knapp S, Kalisz H, Flocco M. 2003. Structural Characterization of the GSK-3? Active Site Using Selective and Non-selective ATP-mimetic Inhibitors. Journal of Molecular Biology. 333(2):393-407. https://doi.org/10.1016/j.jmb.2003.08.031
3.
Cohen P, Goedert M. 2004. GSK3 inhibitors: development and therapeutic potential. Nat Rev Drug Discov. 3(6):479-487. https://doi.org/10.1038/nrd1415
4.
Dajani R, Fraser E, Roe S, Young N, Good V, Dale TC, Pearl LH. 2001. Crystal Structure of Glycogen Synthase Kinase 3?. Cell. 105(6):721-732. https://doi.org/10.1016/s0092-8674(01)00374-9
5.
Doble BW. 2003. GSK-3: tricks of the trade for a multi-tasking kinase. 116(7):1175-1186. https://doi.org/10.1242/jcs.00384
6.
Droucheau E, Primot A, Thomas V, Mattei D, Knockaert M, Richardson C, Sallicandro P, Alano P, Jafarshad A, Barrate B, et al. 2004. Erratum to: ?Plasmodium falciparum glycogen synthase kinase-3: molecular model, expression, intracellular localisation and selective inhibitors? [Biochim. Biophys. Acta 1697 (2004) 181?196]. Biochimica et Biophysica Acta (BBA) - Proteins and Proteomics. 1700(1):139-140. https://doi.org/10.1016/j.bbapap.2004.04.005
7.
Gao C, Hölscher C, Liu Y, Li L. 2012. GSK3: a key target for the development of novel treatments for type 2 diabetes mellitus and Alzheimer disease. 23(1):1-11. https://doi.org/10.1515/rns.2011.061
8.
Gould TD, Zarate CA, Manji HK. 2004. Glycogen Synthase Kinase-3. J. Clin. Psychiatry. 65(1):10-21. https://doi.org/10.4088/jcp.v65n0103
9.
Hur E, Zhou F. 2010. GSK3 signalling in neural development. Nat Rev Neurosci. 11(8):539-551. https://doi.org/10.1038/nrn2870
10.
Jope RS. 2003. Lithium and GSK-3: one inhibitor, two inhibitory actions, multiple outcomes. Trends in Pharmacological Sciences. 24(9):441-443. https://doi.org/10.1016/s0165-6147(03)00206-2
11.
Klein PS, Melton DA. 1996. A molecular mechanism for the effect of lithium on development.. Proceedings of the National Academy of Sciences. 93(16):8455-8459. https://doi.org/10.1073/pnas.93.16.8455
12.
Leclerc S, Garnier M, Hoessel R, Marko D, Bibb JA, Snyder GL, Greengard P, Biernat J, Wu Y, Mandelkow E, et al. 2001. Indirubins Inhibit Glycogen Synthase Kinase-3? and CDK5/P25, Two Protein Kinases Involved in Abnormal Tau Phosphorylation in Alzheimer's Disease. J. Biol. Chem.. 276(1):251-260. https://doi.org/10.1074/jbc.m002466200
13.
MEIJER L, FLAJOLET M, GREENGARD P. 2004. Pharmacological inhibitors of glycogen synthase kinase 3. Trends in Pharmacological Sciences. 25(9):471-480. https://doi.org/10.1016/j.tips.2004.07.006
14.
Palomo V, I. Perez D, Gil C, Martinez A. 2011. The Potential Role of Glycogen Synthase Kinase 3 Inhibitors as Amyotrophic Lateral Sclerosis Pharmacological Therapy. CMC. 18(20):3028-3034. https://doi.org/10.2174/092986711796391697
15.
Phiel CJ, Wilson CA, Lee VM, Klein PS. 2003. GSK-3? regulates production of Alzheimer's disease amyloid-? peptides. Nature. 423(6938):435-439. https://doi.org/10.1038/nature01640
16.
Phukan S, Babu V, Kannoji A, Hariharan R, Balaji V. 2010. GSK3?: role in therapeutic landscape and development of modulators. 160(1):1-19. https://doi.org/10.1111/j.1476-5381.2010.00661.x
17.
Rayasam GV, Tulasi VK, Sodhi R, Davis JA, Ray A. Glycogen synthase kinase 3: more than a namesake. 156(6):885-898. https://doi.org/10.1111/j.1476-5381.2008.00085.x
18.
Sato N, Meijer L, Skaltsounis L, Greengard P, Brivanlou AH. 2004. Maintenance of pluripotency in human and mouse embryonic stem cells through activation of Wnt signaling by a pharmacological GSK-3-specific inhibitor. Nat Med. 10(1):55-63. https://doi.org/10.1038/nm979