MilliporeSigma
HomeWestern BlottingWestern Blot Analysis of Immunoprecipitation (IP-Western)
Western Blot Analysis of Immunoprecipitation (IP-Western)
IP-Western analysis remains a popular technique for identifying protein-protein interactions and identifying unknown proteins in a multi-protein complex. The steps include cell lysis, formation of the antibody-antigen (immune) complex, precipitation of the immune complexes, and analysis by Western blotting.
Read more about
Related Products
Loading
High Background
High background, leading to nonspecific bands on Western blot
Figure 1.High background, leading to nonspecific bands on Western blot.
Possible Cause               Remedy
Non-specific binding to beads
  • Perform a pre-clearing step when using agarose beads. Adding non-coated agarose beads (prior to the addition of antibody-coated beads) to the protein mixture removes proteins that non-specifically bind to the agarose.
  • Ensure beads are properly blocked with BSA prior to adding antibody.
  • Decrease volume of beads.
Wash buffer not stringent enough
  • Switch to higher stringency buffer such as one that is RIPA-based.
Non-specific antibody binding
  • Decrease incubation time, lysate volume/concentration, and/or antibody concentration.
  • Use a more specific antibody.
Inadequate washing
  • Add more washes and/or wash for a longer period of time.
  • Increase stringency of the wash buffer.
  • Invert tube several times to ensure adequate washing.
No Target Protein Eluted, or Not Enough Target Protein Eluted
Possible CauseRemedy
Protein degradation, dephosphorylation, and denaturation during lysis step
  • Perform all steps at 4 °C. Ensure adequate amount of protease and phosphatase inhibitors.
Lysis buffer too stringent
  • Switch to a medium or low-stringency buffer such as one that is NP-40-, Triton® X-100-, or PBS-based.
Antibody not bound to the beads
  • Ensure beads are properly matched to the antibody isotype.
Not enough antibody-antigen binding
  • Increase the concentration of antibody added to the lysate mixture and/or the concentration of beads.
  • Incubate antibody and lysate mixture for a longer time. 
  • In cases of low protein concentration or low antibody-protein affinity, switch from a direct immunoprecipitation method to an indirect method. 
Agarose beads accidentally removed during precipitation step
  • Ensure agarose beads are not aspirated into the pipette.
  • Use magnetic beads instead of agarose beads to minimize/eliminate bead loss.
Mouse IgG or chicken antibody does not bind efficiently to protein G- or protein-A conjugated beads (for indirect immunoprecipitation)
  • Add a bridging antibody during the precipitation step to improve the binding of these antibodies to the protein G- or protein A-conjugated beads.
Target protein remains on the beads
  • Ensure appropriate elution buffer type and pH.
Disruption of protein complexes (in co-immunoprecipitation experiments)
  • Vortex gently and use wide pipette tips to avoid disruption of protein complexes.
Extra Band(s) at 50 kDa and 20 kDa on Final Western Blot that May Mask Band of Interest
In a cross-linking IP western, the heavy and light chains of the primary capture antibody are excluded from the sample.
Figure 2.In a cross-linking IP western, the heavy and light chains of the primary capture antibody are excluded from the sample.
Possible Cause                         Remedy
Heavy and light chains of the primary antibody are being recognized by the secondary antibody
  • Use a secondary antibody that detects native
    (non-denatured) immunoglobulins.
  • Use crosslinking IP by first crosslinking protein A or G beads to the capture antibody using DSS
Guide to Choosing Immunoprecipitation Beads
Relative Affinity
AntibodiesProtein A/G MixProtein AProtein GKappa Ig BinderLambda Ig BinderKappa/Lambda Mix*
Rabbit IgGSASASA   
Mouse IgMRESA    
Mouse IgG3SAMSASA   
Mouse IgG2bSASASA   
Mouse IgG2aSASASA   
Mouse IgG1MSAMSAMSA   
Human IgMMSAMSA MSAMSASA
Human IgEMSAMSA MSAMSASA
Human IgDMSAMSA MSAMSASA
Human IgAMSAMSA MSAMSASA
Human IgG4SASASAMSAMSASA
Human IgG3SA SAMSAMSASA
Human IgG2SASASAMSAMSASA
Human IgG1SASASAMSAMSASA
Rat IgMRERE    
Rat IgG2cMSAREMSA   
Rat IgG2bMSAREMSA   
Rat IgG2aSARESA   
Rat IgG1MSAREMSA   
Rat IgGMSASASA   
FragmentsProtein A/G MixProtein AProtein GKappa Ig BinderLambda Ig BinderKappa/Lambda Mix*
Human l    SAMSA
Human k   SA MSA
Human FcMSAMSAMSA   
Human scFvMSAMSA RERERE
Human F(ab')2MSAMSAMSAMSAMSASA
Human F(ab)MSAMSAMSAMSAMSASA
Key code for relative affinity of protein A and G; PureProteome™ Kappa and Lambda magnetic beads for antibodies: SA = Strong Affinity; MSA = Moderate/Slight Affinity; RE = Requires Evaluation
*PureProteome™ Kappa/Lambda mix is not a catalog item. Simply procure the Kappa and Lambda beads individually and mix at a 1:1 ratio.