Skip to Content
Merck
All Photos(1)

Key Documents

790214

Sigma-Aldrich

Poly(L-lactide-co-glycolide)

L-lactide:glycolide 5:95, viscosity ≥ 1.1 dL/g

Synonym(s):

MAXPRENE® MX2, PLA-co-PGA, PLGA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C3H4O2)m(C2H2O2)n
UNSPSC Code:
12352100
NACRES:
NA.23

Pricing and availability is not currently available.

form

granular

feed ratio

lactide:glycolide 5:95

degradation timeframe

<4 months

viscosity

1.1 dL/g ((0.1% (w/v) in HFIP, 25 °C))

transition temp

Tm 195 °C

solubility

soluble (fluorinated sovents)

storage temp.

2-8°C

Application

This biodegradable PLGA has improved processing over the PLGA achieved using a feed ratio of 10:90 lactide to glycolide for use in multifilament, microfilament, textiles, and molded articles.

Other Notes

Strength Loss: 2-3 weeks depending on construct
Mass Loss: 3-4 months depending on construct
Mechanical Properties:Molded Article (ASTM Type V Tensile Bar)
Yield Point: 421.1 N
Modulus: 2632 MPa
Elongation at break: 4.3 %
Yield Stress: 78.8 MPa
Melt Flow Index: 10.04 g/10 minutes (230 °C with 2160 g weight applied)

Legal Information

Product of Poly-Med, Inc.
Max-Prene is a registered trademark of Poly-Med, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Innovations in polymer technology have had a significant impact on the advancement of novel drug delivery systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service