Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

G0419

Sigma-Aldrich

GR 55562 dihydrobromide

>97% (HPLC), powder

Synonym(s):

3-[3-(Dimethylamino)propyl]-4-hydroxy-N-[4-(4-pyridinyl)phenyl]benzamide dihydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H25N3O2 · 2HBr
CAS Number:
Molecular Weight:
537.29
MDL number:
UNSPSC Code:
12352202

Pricing and availability is not currently available.

description

Store tightly sealed with desiccant.

Assay

>97% (HPLC)

form

powder

color

light yellow

solubility

H2O: ~12 mg/mL (with mixing)

storage temp.

2-8°C

InChI

1S/C23H25N3O2.ClH/c1-26(2)15-3-4-19-16-20(7-10-22(19)27)23(28)25-21-8-5-17(6-9-21)18-11-13-24-14-12-18;/h5-14,16,27H,3-4,15H2,1-2H3,(H,25,28);1H

InChI key

NUDSRNNZTVHCKY-UHFFFAOYSA-N

Biochem/physiol Actions

5-HT1B/5-HT1D serotonin receptor antagonist

Features and Benefits

This compound was developed by GlaxoSmithKline. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Legal Information

Sold for research purposes under agreement from Glaxo­Smith­Kline

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zheng-Ming Ding et al.
Psychopharmacology, 204(3), 381-390 (2009-01-24)
Previous studies indicated that ethanol could be self-infused into the posterior ventral tegmental area (p-VTA) and that activation of local serotonin-3 (5-HT(3)) receptors was involved. 5-HT(1B) and 5-HT(2A) receptors are involved in the effects of 5-HT and ethanol on VTA
Edmund Przegaliński et al.
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology, 14(3), 217-225 (2004-04-02)
The present study was aimed at finding out whether 5-HT(1B) receptors located in the ventral tegmental area (VTA) played a role in the locomotor hyperactivity induced by a single dose of cocaine and in the sensitization evoked by repeated exposure
Iwona Papla et al.
Polish journal of pharmacology, 54(4), 351-357 (2003-01-14)
Previous research demonstrated that the mesoaccumbens dopamine (DA) pathway played a critical role in the behavioral effects of amphetamine in rodents. Nonetheless, recent findings have also indicated involvement of 5-hydroxytryptamine (5-HT, serotonin) transmission in these effects. In the present study
Ji-Hyun Kim et al.
Pharmacology, biochemistry, and behavior, 87(1), 41-47 (2007-05-05)
The present study was undertaken to investigate the antidepressant-like effects of the methylene chloride fraction of Albizzia julibrissin (MCAJ) using a tail suspension test in mice. MCAJ was orally administered at 50, 100, or 200 mg/kg to mice, 1 h
E Przegaliński et al.
Journal of physiology and pharmacology : an official journal of the Polish Physiological Society, 53(3), 383-394 (2002-10-09)
The present study was designed to examine the effect of 5-HT1B receptor ligands microinjected into the subregions of the nucleus accumbens (the shell and the core) on the locomotor hyperactivity induced by cocaine in rats. Male Wistar rats were implanted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service