Skip to Content
Merck
All Photos(1)

Key Documents

C1490

SAFC

EX-CELL® CD CHO-3 Medium, Chemically Defined

Animal-component free, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75

Pricing and availability is not currently available.

description

for research or for further manufacturing use

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

components

L-glutamine: no
HEPES: yes

shipped in

ambient

storage temp.

2-8°C

Application

Complete, chemically-defined medium formulated to optimize cell growth and protein expression in Chinese hamster ovary (CHO) cells.

Features and Benefits

Developed to meet the needs of biotechnology and vaccine manufacturing, this medium supports rapid initial cell growth and high levels of protein expression in suspension cultures. This medium supports and maintains high cell densities for extended periods, resulting in increased productivity.

Other Notes

Proprietary formulation containing inorganic salts, HEPES and sodium bicarbonate buffers, essential and non-essential amino acids, vitamins, recombinant human insulin, other organic compounds, trace elements, and surfactants.
Does not contain antibiotics, antimycotics, L-glutamine, transferrin, hydrolysates, or other undefined nutrients or supplements. Contains no animal-derived components or components synthesized from animal-derived materials.

Caution

Do not freeze. Protect from light. Several components of the medium are light labile and should not be exposed to light for lengthy periods of time.

Reconstitution

Aseptically add 20-40 mL of 200 mM L-glutamine solution per liter of medium prior to use.

Legal Information

EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

A signal peptide is a 5-30 amino acid (aa) peptide present at the N-terminus of secretory proteins.

In the present study, we have identifi ed species-specifi c housekeeping genes (HKGs) for Chinese Hamster Ovary (CHO) cells using data from microarray gene expression profiling.

MGAT1 adds N-acetylglucosamine to the Man5GlcNAc2 (Man5) structure. Goh et al. reported increased sialylation after restoring MGAT1 function in MGAT1 deficient CHO cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service