Merck
50688-U
Supelco
Ascentis® Express 90 Å PCS-Phenyl-Hexyl, 2.7 µm HPLC Column
column L × I.D. 100 mm × 1.5 mm, pk of 1 ea
ログイン組織・契約価格を表示する
About This Item
UNSPSCコード:
41115709
NACRES:
SB.52
カラムL × 内径:
100 mm × 1.5 mm
包装
pk of 1 ea
品質水準
カラムL × 内径
100 mm × 1.5 mm
類似した製品をお探しですか? 訪問 製品比較ガイド
詳細
Ascentis® Express PCS-Phenyl-Hexyl columns are specifically engineered to provide efficient separation of basic, acidic, or neutral compounds under low ionic strength (formic acid) mobile phase conditions. These columns contain advanced Fused-Core® particles with pores strategically sized to 90 Å for small molecule analyses. This attribute makes these columns an excellent choice for pharmaceutical method development. The unique PCS-C18 chemistry enables exceptional peak shape and enhanced loading capacity of basic compounds as compared to traditional C18 chemistries. This column also provides orthogonal selectivity to other small molecule chemistries.
アナリシスノート
The target analytes include basic compounds.
法的情報
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany
Fused-Core is a registered trademark of Advanced Materials Technology, Inc.
最新バージョンのいずれかを選択してください:
試験成績書(COA)